Skip to Content
Merck
All Photos(3)

Key Documents

203939

Sigma-Aldrich

Palladium

powder, 99.995% trace metals basis

Synonym(s):

Palladium (powder), Palladium black, Palladium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.995% trace metals basis

form

powder

resistivity

9.96 μΩ-cm, 20°C

particle size

0.25-2.36 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Palladium(Pd) is a silver-white noble transition metal in the platinum group of metals. Relative to the platinum group metals, Pd has the lowest density, 12 g/cm3, and lowest melting point, 1555 °C. Pd is known for its strong catalytic activity and is used in various applications such as electrical equipment, dental materials, implanted medical devices, and automobile catalysts. Additionally, it is also extensively employed in the preparation of diverse heterogeneous palladium catalysts for various organic transformations. Our high-purityPd powder is differentiated by its high trace metals purity, which makes it particularly useful in sensitive research projects in electronics, catalysis, and synthesis.

Application


  • Advances in silicon-carbon composites anodes derived from agro wastes for applications in lithium-ion battery: A review.: This review highlights the integration of graphite with silicon derived from agricultural waste to create high-performance anodes for lithium-ion batteries, underscoring the importance of sustainable material sources in battery technology (Fafure et al., 2024).

  • Edge-enhanced super microgenerator based on a two-dimensional Schottky junction.: Graphite′s electrical properties are leveraged to enhance the efficiency of microgenerators, contributing significantly to advancements in micro-electromechanical systems (MEMS) and energy harvesting technologies (Yu et al., 2024).

  • Batch and semi-continuous treatment of cassava wastewater using microbial fuel cells and metataxonomic analysis.: Demonstrates graphite′s application in microbial fuel cells for the treatment of wastewater, providing an environmentally friendly solution for waste management and energy generation in industrial processes (Quintero-Díaz and Gil-Posada, 2024).


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Volarevic et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 18(1), 131-137 (2013-04-25)
As novel therapeutic agents relevant to colon cancer therapy are explored continuously, we tested 4 R2edda-type ligand precursors O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)pentanoic acid (L1.2HCl-L4.2HCl) and corresponding palladium(II) and platinum(II) complexes against the human colon cancer cell lines CaCo-2, SW480 and
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work
Stefano Nicolai et al.
The Journal of organic chemistry, 78(8), 3783-3801 (2013-03-21)
Tetrahydrofurans and pyrrolidines are among the most important heterocycles found in bioactive compounds. Cyclization-functionalization domino reactions of alcohols or amines onto olefins constitute one of the most efficient methods to access them. In this context, oxy- and aminoalkynylation are especially
Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Daniel Smykowski et al.
Journal of molecular graphics & modelling, 41, 89-96 (2013-03-21)
This study is the analysis of the adsorption process of the CO2 molecule on the cationic sites of the DOH zeolite. Based on the DFT method, we have been able to identify several adsorption sites containing extra-framework cations and evaluate

Articles

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service