Merck
  • Magnetic graphene solid-phase extraction for the determination of carbamate pesticides in tomatoes coupled with high performance liquid chromatography.
Magnetic graphene solid-phase extraction for the determination of carbamate pesticides in tomatoes coupled with high performance liquid chromatography.
Talanta (2015-05-13)
Na Li, Juan Chen, Yan-Ping Shi
ABSTRACT
Graphene-based magnetic nanoparticles, comprising zero-valent iron, iron oxide-oxyhydroxide and graphene, were prepared through a simple one-step synthesis method, and subsequently applied to magnetic solid-phase extraction for the determination of trace carbamate pesticides in tomatoes coupled with high performance liquid chromatography. The properties of the nanocomposites were confirmed by using Fourier transform infrared spectroscopy, X-ray photoelectron spectroscopy, and vibrating sample magnetometer. The components within the nanocomposites endowed the material with high extraction performance and manipulative convenience. Compared with reduced graphene oxide, the as-prepared G-MNPs showed the better extraction efficiencies for the carbamate pesticides thanks to the contribution of the iron-containing magnetic nanoparticles to the adsorption capacity of the nanocomposites. Various experimental parameters affecting the extraction efficiency had been investigated in detail. Under the optimal conditions, the method provided high enrichment factors ranging from 364 to 434, good linearities ranging from 5 to 200ng g(-1) for metolcarb, baygon and methiocarb and 10 to 200ng g(-1) for carbofuran and isoprocarb, low limits of detection ranging from 0.58 to 2.06ng g(-1), and satisfactory spiked recoveries (between 90.34% and 101.98% with the relative standard deviation values from 1.21% to 5.93%). It was confirmed that this novel method was an efficient pretreatment and enrichment procedure and could be successfully applied for extraction and determination of trace carbamate pesticides in complex matrices.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Carbofuran, 98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis