Merck
  • Subcellular mRNA Localization Regulates Ribosome Biogenesis in Migrating Cells.
Subcellular mRNA Localization Regulates Ribosome Biogenesis in Migrating Cells.
Developmental cell (2020-11-11)
Maria Dermit, Martin Dodel, Flora C Y Lee, Muhammad S Azman, Hagen Schwenzer, J Louise Jones, Sarah P Blagden, Jernej Ule, Faraz K Mardakheh
ABSTRACT
Translation of ribosomal protein-coding mRNAs (RP-mRNAs) constitutes a key step in ribosome biogenesis, but the mechanisms that modulate RP-mRNA translation in coordination with other cellular processes are poorly defined. Here, we show that subcellular localization of RP-mRNAs acts as a key regulator of their translation during cell migration. As cells migrate into their surroundings, RP-mRNAs localize to the actin-rich cell protrusions. This localization is mediated by La-related protein 6 (LARP6), an RNA-binding protein that is enriched in protrusions. Protrusions act as hotspots of translation for RP-mRNAs, enhancing RP synthesis, ribosome biogenesis, and the overall protein synthesis in migratory cells. In human breast carcinomas, epithelial-to-mesenchymal transition (EMT) upregulates LARP6 expression to enhance protein synthesis and support invasive growth. Our findings reveal LARP6-mediated mRNA localization as a key regulator of ribosome biogenesis during cell migration and demonstrate a role for this process in cancer progression downstream of EMT.
Sigma-Aldrich
MISSION® esiRNA, targeting human LARP1
Sigma-Aldrich
Emetine dihydrochloride
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
MISSION® siRNA Product Offerings, Custom and Predesigned siRNA
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
Homoharringtonine, ≥98% (HPLC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%