Skip to Content
Merck
All Photos(3)

Key Documents

C6158

Sigma-Aldrich

Crystal Violet

≥90% purity anhydrous basis, ACS reagent, powder

Synonym(s):

Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
Colour Index Number:
42555
Beilstein:
3580948
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Crystal Violet, ACS reagent, ≥90.0% anhydrous basis

grade

ACS reagent

Quality Level

Assay

≥90.0% anhydrous basis

form

powder

loss

≤7.5% loss on drying

color

green to very dark green

pH

2.5-3.5 (20 °C, 10 g/L)

mp

205 °C (dec.) (lit.)

solubility

water: soluble 50 g/L at 27 °C

density

1.19 g/cm3 at 20 °C

absorption

passes test

suitability

passes test for sensitivity as indicator

antibiotic activity spectrum

fungi

application(s)

diagnostic assay manufacturing
hematology
histology

Mode of action

cell membrane | interferes
enzyme | inhibits

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Crystal violet has been used to determine cell viability. It has been used in the staining of cells for the quantification of cellular migration.

Biochem/physiol Actions

Crystal violet can be used for DNA visualization in agarose gels. The dye is used only in the presence of high concentrations of DNA. Crystal violet is also used for the staining of bacteria in gram staining technique. It is also used for the staining of plant chromosomes. Crystal violet helps in colorimetric measurement of cell viability.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulation of survivin expression exerts antitumoral effects on mouse breast cancer cells in vitro and in vivo.
Ma WH, et al.
Oncology Letters, 11, 159-159 (2016)
The activation of c-Jun NH2-terminal kinase is required for dihydroartemisinin-induced autophagy in pancreatic cancer cells.
Jia G, et al.
Journal of Experimental & Clinical Cancer Research, 33, 8-8 (2014)
Plant Chromosomes (1999)
Carson S and Robertson D
Manipulation and Expression of Recombinant DNA (2005)
Eric H Clifton et al.
PloS one, 13(3), e0194815-e0194815 (2018-03-23)
Terrestrial plants can harbor endophytic fungi that may induce changes in plant physiology that in turn affect interactions with herbivorous insects. We evaluated whether the application of entomopathogenic fungi Beauveria bassiana and Metarhizium brunneum to soybean seeds could become endophytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service