Merck
  • Probing hot spots at protein-ligand binding sites: a fragment-based approach using biophysical methods.
Probing hot spots at protein-ligand binding sites: a fragment-based approach using biophysical methods.
Journal of medicinal chemistry (2006-08-04)
Alessio Ciulli, Glyn Williams, Alison G Smith, Tom L Blundell, Chris Abell
ABSTRACT
Mapping interactions at protein-ligand binding sites is an important aspect of understanding many biological reactions and a key part of drug design. In this paper, we have used a fragment-based approach to probe "hot spots" at the cofactor-binding site of a model dehydrogenase, Escherichia coli ketopantoate reductase. Our strategy involved the breaking down of NADPH (Kd = 300 nM) into smaller fragments and the biophysical characterization of their binding using WaterLOGSY NMR spectroscopy, isothermal titration calorimetry (ITC), and inhibition studies. The weak binding affinities of fragments were measured by direct ITC titrations under low c value conditions. The 2'-phosphate and the reduced nicotinamide groups were found to contribute a large part of the binding energy. A combination of ITC and site-directed mutagenesis enabled us to locate the fragments at separate hot spots on opposite ends of the cofactor-binding site. This study has identified structural determinants for cofactor recognition that represent a blueprint for future inhibitor design.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis