Skip to Content
Merck
All Photos(1)

Key Documents

438081

Sigma-Aldrich

Phosphoric acid

greener alternative

ACS reagent, ≥85 wt. % in H2O

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

Assay

≥85% (with NaOH, titration)

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

≥85 wt. % in H2O

technique(s)

NMR: suitable

impurities

Reducing substances, passes test
≤0.001% volatile acids (as CH3COOH)

color

APHA: ≤10

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤3 ppm
nitrate (NO3-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

As: ≤1 ppm
Ca: ≤0.002%
Fe: ≤0.003%
K: ≤0.005%
Mg: ≤0.02%
Mn: ≤0.5 ppm
Na: ≤0.025%
Sb: ≤0.002%
heavy metals: ≤0.001% (by ICP)

greener alternative category

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Phosphoric acid (H3PO4), also known as orthophosphoric acid, is an inorganic acid. It has a wide range of industrial applications such as an intermediate to produce animal feedstock, to prepare stock solutions for workup procedures and to treat the metal surfaces for better adhesion of paint and resin coatings.[1][2] In the food industry it is used as a preservative, an acidulant and flavor enhancer.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Phosphoric acid may be employed as standard for the determination of phosphorus-31 nuclear magnetic resonance (31P NMR) chemical shifts of the common phospholipids.[3] It may be employed in the following studies:
  • As external standard for the analysis of lipid composition of human white matter by 31P NMR spectroscopy.[4]
  • As external standard during the acid-labile phosphorylation of histone H4 by 31P NMR spectroscopy.[5]
  • As reference standard to investigate the surface species present on silica/alumina-supported sodium phosphates by pre- and postreaction MAS31P-NMR and FTIR spectroscopies.[6]
  • As catalyst in the vapor-phase Fries rearrangement of phenylacetate at 623K over a series of surface modified and unmodified ZSM-5.[7]
  • As an eco-friendly reagent for the removal of protecting groups like tert-butyl carbamates, tert-butyl esters and tert-butyl ethers.[8]

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Myers RL.
The 100 Most Important Chemical Compounds: A Reference Guide, 220-220 (2007)
A simple method of increasing the adhesion of acrylic filling materials to enamel surfaces.
Buonocore MG.
Journal of Dental Research, 34(6), 849-853 (1955)
Aqueous phosphoric acid as a mild reagent for deprotection of tert-butyl carbamates, esters, and ethers.
Li B, et al.
The Journal of Organic Chemistry, 71(24), 9045-9050 (2006)
FTIR and 31 P-NMR spectroscopic analyses of surface species in phosphate-catalyzed lactic acid conversion.
Gunter GC, et al.
J. Catal., 164(1), 207-219 (1996)
P Meneses et al.
Journal of lipid research, 29(5), 679-689 (1988-05-01)
The common phospholipids from biological sources were quantitated using phosphorus-31 nuclear magnetic resonance (NMR) spectroscopy in conjunction with an analytical reagent composed of two parts: 1) 2 ml of reagent chloroform in which was dissolved 0.01-100 mg of crude tissue

Protocols

To measure alcohol dehydrogenase activity, this assay uses β-nicotinamide adenine dinucleotide phosphate and a continuous spectrophotometric rate determination at 340 nm.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service