Merck
  • Nerve growth factor upregulates sirtuin 1 expression in cholestasis: a potential therapeutic target.
Nerve growth factor upregulates sirtuin 1 expression in cholestasis: a potential therapeutic target.
Experimental & molecular medicine (2018-01-13)
Ming-Shian Tsai, Po-Huang Lee, Cheuk-Kwan Sun, Ting-Chia Chiu, Yu-Chun Lin, I-Wei Chang, Po-Han Chen, Ying-Hsien Kao
ABSTRACT
This study investigated the regulatory role of nerve growth factor (NGF) in sirtuin 1 (SIRT1) expression in cholestatic livers. We evaluated the expression of NGF and its cognate receptors in human livers with hepatolithiasis and the effects of NGF therapy on liver injury and hepatic SIRT1 expression in a bile duct ligation (BDL) mouse model. Histopathological and molecular analyses showed that the hepatocytes of human diseased livers expressed NGF, proNGF (a precursor of NGF), TrkA and p75NTR, whereas only p75NTR was upregulated in hepatolithiasis, compared with non-hepatolithiasis livers. In the BDL model without NGF therapy, p75NTR, but not TrkA antagonism, significantly deteriorated BDL-induced liver injury. By contrast, the hepatoprotective effect of NGF was abrogated only by TrkA and not by p75NTR antagonism in animals receiving NGF therapy. Intriguingly, a positive correlation between hepatic SIRT1 and NGF expression was found in human livers. In vitro studies demonstrated that NGF upregulated SIRT1 expression in mouse livers and human Huh-7 and rodent hepatocytes. Both NGF and proNGF induced protective effects against hydrogen peroxide-induced cytotoxicity in Huh-7 cells, whereas inhibition of TrkA and p75NTR activity prevented oxidative cell death. Mechanistically, NGF, but not proNGF, upregulated SIRT1 expression in human Huh-7 and rodent hepatocytes via nuclear factor (NF)-κB activity, whereas NGF-induced phosphoinositide-3 kinase/Akt, extracellular signal-regulated kinase and NF-κB signaling and SIRT1 activity were involved in its hepatoprotective effects against oxidative injury. These findings suggest that pharmacological manipulation of the NGF/SIRT1 axis might serve as a novel approach for the treatment of cholestatic disease.
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
EX-527, ≥98% (HPLC)
Sigma-Aldrich
Anti-Nerve Growth Factor-β Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Nerve Growth Factor-β Antibody, Chemicon®, from sheep