Skip to Content
Merck
All Photos(4)

Key Documents

MAB1501

Sigma-Aldrich

Anti-α-Actin-1 (ACTA1) antibody

CHEMICON®, mouse monoclonal, C4

Synonym(s):

MAB1501X, MAB1501R, Pan Actin antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

C4, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Actins are ubiquitous eukaryotic proteins that serve as a multi-functional, basic building blocks of cytoskeletal microfilaments. They play critical roles in a wide range of cellular processes, including cell division, cell migration, chromatin remodeling, trascriptional regulation and vesicle trafficking. These funstions are attributed to their ability to form filaments, which can quickly assemble and disassemble depending upon the needs of the cell. At least six different actin types have been reported in mammals. Although actins show about 90% overall sequence homology, isoforms do not show spatial, temporal and tissue-specific expression patterns and only 50-60% homology is found in their 18 N-terminal residues. Beta and gamma-actins, also known as cytoplasmic actins, are highly conserved in higher animals and are predominantly expressed in non-muscle cells where they control cell structure. Exocytosis, and motility. They are nearly identical proteins and differ only in four amino acids at the N-terminal region. The other four actin isoforms are typically found in specific adult muscle tissue types. Alpha-cardiac and alpha-skeletal actins are expressed in striated cardiac and skeletal muscles, respectively. Alpha and gamma actins are primarily found in vascular smooth muscle and enteric smooth muscles, respectively. It has been shown that under calcium-bound conditions, beta-actin exhibits more dynamic behavior than gamma-actin with higher rates of polymerization and depolymerization. Also, beta- and gamma-actins can readily copolymerize, and the resulting filaments exhibit polymerization and depolymerization rates that vary depending on the ration of beta- to gamma-actin (Lessard, JL.,et al.(1988). Cell Motility Cytoskeleton 10(3); 349-362.

Specificity

MAB1501 is a pan-actin antibody that binds to an epitope in a highly conserved region of actin; therefore, this antibody reacts with all six isoforms of vertabrate actin (Lessard, 1988). Reacts with both globular (G) and fillimentous (F) forms of actin and does not interfere with actin polymerization to form filaments, at a ratio as high as one antibody per two actin monomers. However, this antibody does increase the extent of polymerization when used at a lower ratio of antibody to actin. In addition to labeling myotubes, anti-actin stains myoblasts and fibroblasts. Although clone C4 is prepared as an antibody to chicken gizzard muscles actin, it reacts with actins from all vertebrates, as well as with Dictyostelium discoideum and Physarum polycephalum actins (Lessard, 1988).
To date, all animal species and cell types with an actin form react by indirect immunofluorescence or immunoblot, including plant actin.

Immunogen

Epitope is conserved in all known actins.
Purified chicken gizzard actin (Lessard, 1988).

Application

Indirect immunofluorescence at 1:100:
Tissue culture cells -- fix with formaldehyde, treat with methanol or acetone.
Glycerinated myofibrils -- fix fibers with formaldehyde, treat with cold methanol. Stains I-bands intensely and stress fibers in human fibroblasts.
Cryostat sections (6 µm) -- quick frozen in isopentane, slides treated with gelatin and formaldehyde.

Immunoblots:
1:100-1:1,000 (Otey, 1987):On muscle homogenates subject to SDS-PAGE, reacts relatively uniformly with a 43 kD protein present in skeletal, cardiac, gizzard and aorta tissues. Appears to react with all isoforms of actin found in these preparations and shows a strong reaction with the alpha-actin found in skeletal, cardiac, and arterial muscle.

Iodination (Lessard, 1979).

Solid phase binding assay ELISA:
1:800-1:1,000 dilution from a previous lot was shown to be strongly reactive with cytoplasmic actin and shows a significant binding to gizzard, skeletal, arterial and cardiac actins. Also shows a significant binding to both Dictyostelium discoidum and Physarum polycephalum.

ELISA:
strongly reactive with the cytoplasmic actin and shows a significant binding to gizzard, skeletal, arterial and cardiac actins. Also shows a significant binding to both Dictyostelium discoidum and Physarum polycephalum.

Optimal working dilutions must be determined by end user.
Reliably and specifically detect actin using this Anti-Actin Antibody, clone C4. This highly published monoclonal antibody is validated for use in ELISA, IC, IF, IH, IH(P) & WB. This mAb is also available as a fluorescent conjugate.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by Western Blot on A431 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected ACTIN on 10 ug of A431 lysates.

Target description

43 kDa

Linkage

Replaces: 04-1040

Physical form

Mouse monoclonal Ascites fluid, with 0.01% sodium azide.
Unpurified

Storage and Stability

Stable at -20°C in undiluted aliquots for up to 12 months from date of receipt. Do not store in a diluted format. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
HeLa whole cell lysate.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of lysosomal membrane protein depletion on the Salmonella-containing vacuole.
Roark, EA; Haldar, K
Testing null
Frontiers in neurology and neuroscience research null
Beta-catenin overexpression in the mouse brain phenocopies lithium-sensitive behaviors.
Gould, TD; Einat, H; O'Donnell, KC; Picchini, AM; Schloesser, RJ; Manji, HK
Neuropsychopharmacology null
Clinical light exposure, photoreceptor degeneration, and AP-1 activation: a cell death or cell survival signal in the rhodopsin mutant retina?
Gu, D; Beltran, WA; Li, Z; Acland, GM; Aguirre, GD
Investigative Ophthalmology & Visual Science null
Oncogenic viral protein HPV E7 up-regulates the SIRT1 longevity protein in human cervical cancer cells.
Allison, SJ; Jiang, M; Milner, J
Aging null

Articles

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Immunofluorescence uses antibody-conjugated fluorescent molecules for protein localization, modification confirmation, and protein complex visualization.

Protocols

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Tips and troubleshooting for FFPE and frozen tissue immunohistochemistry (IHC) protocols using both brightfield analysis of chromogenic detection and fluorescent microscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service