Merck
  • In Vitro and In Vivo Evaluations of Mesoporous Iron Particles for Iron Bioavailability.
In Vitro and In Vivo Evaluations of Mesoporous Iron Particles for Iron Bioavailability.
International journal of molecular sciences (2019-10-28)
Jung-Feng Lin, Chau-Chung Wu, Yu-Jiun Liao, Subhaini Jakfar, Zi-Biao Tang, Jhewn-Kuang Chen, Feng-Huei Lin
ABSTRACT
Chronic renal failure involving hemodialysis results in blood loss during filtration. Iron deficiency and iron deficiency anemia can result. A compensatory increase in iron dosage has many side effects including discomfort. Elemental iron is a highly-pure iron source, which reduces the frequency of dosages; the solubility decreases with increased particle size or pore size. In this study, synthesized mesoporous iron particles (MIPs) were used to relieve iron deficiency anemia. Their bioavailability was measured in vitro by a Caco-2 cell model and in vivo in iron-deficient rats. In vitro bioavailability of MIPs was examined by measuring ferritin content in the Caco-2 cell model. Iron uptake of MIPs was significantly higher than commercial iron particles, which were less porous. In vivo bioavailability of MIPs was examined by measuring body weight gain and red blood cell-related parameters, compared with the bioavailability of standard drug ferrous sulfate in iron-deficient anemic rats. Finally, average hemoglobin content and hemoglobin regeneration efficiency were significantly higher in anemic rats supplemented with commercial iron particles, compared to anemic controls. In the 28-day oral toxicity test, MIPs were not significantly toxic to rat physiology or tissue histopathology. Thus, MIPs may allow effective recovery of hemoglobin in iron deficiency anemia.
Sigma-Aldrich
holo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
ECO TWEEN® 80, viscous liquid
Sigma-Aldrich
Iron(III) chloride, reagent grade, 97%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Pancreatin from porcine pancreas, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, γ-irradiated, powder, suitable for, suitable for cell culture
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
TWEEN® 80, viscous liquid