Skip to Content
Merck
All Photos(1)

Key Documents

A5955

Sigma-Aldrich

Antibiotic Antimycotic Solution (100×), Stabilized

suspension, suitable for cell culture, BioReagent

Synonym(s):

Penicillin-Streptomycin-Amphotericin B Suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.76
Pricing and availability is not currently available.

Product Name

Antibiotic Antimycotic Solution (100×), Stabilized, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture

Quality Level

sterility

0.1 μm filtered

product line

BioReagent

form

suspension

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
yeast

application(s)

agriculture

Mode of action

cell wall synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Antibiotic Antimycotic Solution is formulated to contain 10,000 units penicillin, 10 mg streptomycin and 25μg Amphotericin B per mL, and is solubilized in a proprietary citrate buffer. It is recommended for use in cell culture applications at 10mL/L. Antibioticantimycotic solution is generally used to prevent the growth of harmful microorganisms including bacteria and fungi in cell culture media.[1]

Application

Antibiotic antimycotic solution has been used:
  • as a supplement in different types of cell culture media.[2][3][4]
  • as a component of phosphate buffer saline (PBS) sterile solution to prevent contamination of periodontal ligament fibroblasts (PDLF) tissues that are isolated and cultured in the experiments.[5]
  • as a component of storage solution for preserving tracheal segments harvested for subsequent experimental investigation.[6]

Biochem/physiol Actions

Mode of Action: Penicillin acts by inhibiting bacterial cell-wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome and causes miscoding. Amphotericin B interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.

Antimicrobial spectrum: Includes Gram-negative bacteria, Gram-positive bacteria, fungi and yeasts.

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of biocide addition on plantlet growth and contamination occurrence during the in vitro culture of blueberry
Sun YH, et al.
Journal of plant biotechnology, 42(2) (2015)
Evaluation of Structural Viability of Porcine Tracheal Scaffolds after 3 and 6 Months of Storage under Three Different Protocols
Bioengineering (Basel, Switzerland), 10(5) (2023)
Monireh Kouhi et al.
Journal of biomedical materials research. Part A, 107(6), 1154-1165 (2019-01-13)
Guided bone regeneration (GBR) has been established to be an effective method for the repair of defective tissues, which is based on isolating bone defects with a barrier membrane for faster tissue reconstruction. The aim of the present study is
Alessandro Taloni et al.
Scientific reports, 7(1), 3748-3748 (2017-06-18)
Classification of morphological features in biological samples is usually performed by a trained eye but the increasing amount of available digital images calls for semi-automatic classification techniques. Here we explore this possibility in the context of acrosome morphological analysis during
Kinue Aihara et al.
Kidney international, 64(4), 1283-1291 (2003-09-13)
Crystal formation and retention are critical events for the formation of kidney stones. Oxalate and calcium oxalate (CaOx) crystals are injurious to renal epithelium, and membranes of injured cells promote crystal adherence and retention. Calcium phosphate (CaP) is the most

Articles

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Related Content

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

See All

Questions

  1. I am looking at the SDS for product number A5955, Antibiotic Antimycotic soln. I do not see Amphotericin B in composition of ingredients (3.2 mixtures). I want to know why. Thank you.

    1 answer
    1. Section 3 of SDS shows only the hazardous component information. Product# A5955 contains microgram amounts of Amphotericin B. At this concentration it is non-hazardous, hence section 3 of SDS does not list Amphotericin B.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service