Skip to Content
Merck
All Photos(1)

Key Documents

1356665

USP

Ketorolac Tromethamine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketorolac

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

InChI

1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2

InChI key

BWHLPLXXIDYSNW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketorolac Tromethamine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Matteo Morotti et al.
The journal of sexual medicine, 11(6), 1495-1504 (2014-03-19)
Voluntary termination of pregnancy (TOP) is a social issue; however, even if it is one of the most common procedures performed in the world, few studies evaluated sexual function changes after medical or surgical TOP. The aim of this study
Animesh Kumar et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 70, 140-151 (2015-02-14)
Migraine is a chronic disorder characterized by significant headache and various associated symptoms which worsen with exertion. Zolmitriptan approved for use in the acute treatment of migraine and related vascular headaches but are limited by high pain recurrence due to
Corinne N Riggin et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(11), 1512-1519 (2014-07-02)
Administration of intra-articular medications, including corticosteroids and analgesics, is common clinical practice for knee pathology and dysfunction. Non-steroidal anti-inflammatory drugs (NSAIDs) are another category of medication commonly prescribed for their analgesic and anti-inflammatory properties. Recent studies demonstrated the efficacy of
Daniel D Singer et al.
Cornea, 34(5), 541-543 (2015-04-01)
Topical nonsteroidal antiinflammatory drugs (NSAIDs) are administered topically for a variety of ophthalmologic conditions. Brand diclofenac and brand ketorolac were previously shown to have topical anesthetic effects in addition to analgesic effects. Using the same method, we measured similar anesthetic
Lingmin He et al.
American journal of ophthalmology, 157(6), 1170-1178 (2014-02-25)
To compare the clinical outcomes of wavefront-guided and wavefront-optimized laser in situ keratomileusis (LASIK). Prospective, randomized, fellow-eye-controlled study. The setting was a single academic institution. The study population included 110 eyes of 55 patients with myopia with and without astigmatism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service