Skip to Content
Merck
All Photos(4)

Documents

I4883

Sigma-Aldrich

Ibuprofen

≥98% (GC), powder, COX inhibitor

Synonym(s):

2-(4-Isobutylphenyl)propanoic acid, Brufen, Motrin, Rebugen, α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Ibuprofen, ≥98% (GC)

biological source

synthetic (organic)

Quality Level

Assay

≥98% (GC)

form

powder

mp

77-78  °C

solubility

ethanol: 50 mg/mL, clear, colorless to faintly yellow

SMILES string

CC(C)Cc1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)

InChI key

HEFNNWSXXWATRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ibuprofen is a commonly used nonsteroidal anti-inflammatory drug (NSAID) known for its pain-relieving, anti-inflammatory, and fever-reducing properties. It can hinder the effectiveness of various antihypertensive medications, including β-adrenergic blockers, angiotensin-converting enzyme inhibitors, angiotensin receptor blockers, and diuretics. Ibuprofen exists in an unbound state in cerebrospinal fluid and is accumulated in the synovial fluid of inflamed joints in arthritis patients.

Application

Ibuprofen has been used:
  • to study its vascular and pulmonary effects on neonatal lung development
  • to study its effects on cell apoptosis, cell proliferation, and histology changes in human cholangiocarcinoma cell lines
  • in the preparation of a terpene-based therapeutic deep eutectic system (THEDES) to investigate its physicochemical, antimicrobial, and anticancer properties

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Effects of Ibuprofen, Naproxen and Diclofenac on cell Apoptosis, Cell Proliferation and Histology Changes in Human Cholangiocarcinoma Cell Lines
Leksomboon R, et al.
Asian Pacific Journal of Cancer Prevention (2022)
Untangling the bioactive properties of therapeutic deep eutectic solvents based on natural terpenes
Silva E, et al.
Current research in medicine (2021)
PharmGKB summary: ibuprofen pathways
Mazaleuskaya LL, et al.
Pharmacogenetics and Genomics (2015)
Vascular and pulmonary effects of ibuprofen on neonatal lung development
Chen X, et al.
Respiratory Research (2023)
Mateusz P Czub et al.
Journal of medicinal chemistry, 63(13), 6847-6862 (2020-05-30)
Every day, hundreds of millions of people worldwide take nonsteroidal anti-inflammatory drugs (NSAIDs), often in conjunction with multiple other medications. In the bloodstream, NSAIDs are mostly bound to serum albumin (SA). We report the crystal structures of equine serum albumin

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service