Skip to Content
Merck
All Photos(1)

Key Documents

1176506

USP

Dexamethasone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dexamethasone 21-acetate, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate, 9α-Fluoro-16α-methylprednisolone-21-acetate, Prednisolone F acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Beilstein:
2342608
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

dexamethasone

form

powder

packaging

pkg of 200 mg

manufacturer/tradename

USP

color

white

mp

460.4-464 °F (238—240°C)

solubility

acetone: freely soluble
dichloromethane: slightly soluble
methanol: freely soluble
water: practically insoluble

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COC(C)=O

InChI

1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1

InChI key

AKUJBENLRBOFTD-RPRRAYFGSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone acetate (Decadron-LA) belongs to the class of long-acting glucocorticoids,[1] commonly used as a pharmaceutical ingredient in creams,[2] ointments,[3] microemulsions, lotions, aerosols, etc.[4]
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Betamethasone Acetate
  • Ciprofloxacin and Dexamethasone Otic Suspension
  • Dexamethasone
  • Dexamethasone Acetate
  • Dexamethasone Acetate Injectable Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dexamethasone Acetate
Pharmacopeia, US
United States Pharmacopeia/National Formulary, 29(5), 1815-1815 (2020)
Textbook of Veterinary Internal Medicine, 1(the), 87-92 (2009)
Development and validation of HPLC method for analysis of dexamethasone acetate in microemulsions.
Urban MCC, et al.
Brazilian Journal of Pharmaceutical Sciences , 45(1), 87-92 (2009)
Lj Zivanovic et al.
Journal of chromatography. A, 1088(1-2), 182-186 (2005-09-01)
In this paper, there was developed a sensitive, precise and accurate reversed-phase liquid chromatographic (RP-HPLC) method and validated for simultaneous determination of lidocaine hydrochloride, dexamethasone acetate (DA) and calcium dobesilate (CD) in suppositories and ointment. Also there was achieved a
Determination of dexamethasone acetate in cream by HPLC.
Garcia CV, et al.
Journal of Pharmaceutical and Biomedical Analysis, 31(3), 597-600 (2003)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service