Skip to Content
Merck
All Photos(1)

Key Documents

Y0001477

Dexamethasone sodium phosphate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-phosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28FNa2O8P
CAS Number:
Molecular Weight:
516.40
Beilstein:
6473066
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dexamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O

InChI

1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1

InChI key

PLCQGRYPOISRTQ-FCJDYXGNSA-L

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone sodium phosphate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hüseyin Simavli et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(8), 650-656 (2014-07-02)
To evaluate the inhibitory effects of propranolol, a nonselective and lipophilic β-adrenergic receptor blocker, on alkali-induced corneal neovascularization (NV). Corneal NV was induced in 24 eyes of 24 Wistar rats using NaOH. Following alkali burn, animals were randomized into 4
Soon Hyung Park et al.
The Laryngoscope, 124(6), 1444-1451 (2013-10-25)
We investigated whether the round window membrane (RWM) vibration can facilitate dexamethasone perfusion via the RWM in patients with sudden hearing loss. Prospective study. We first performed an in vitro study using a semipermeable membrane. In the subsequent in vivo
Marcus Ang et al.
The British journal of ophthalmology, 98(8), 1028-1032 (2014-03-29)
To describe outcomes and complications following Descemet's stripping automated endothelial keratoplasty (DSAEK) in eyes with pseudophakic bullous keratopathy (BK) while retaining the anterior chamber intraocular lenses (ACIOL). We included consecutive patients who underwent DSAEK for BK at a single tertiary
Yakov Goldich et al.
American journal of ophthalmology, 159(1), 155-159 (2014-12-03)
To compare objective and subjective outcomes after Descemet membrane endothelial keratoplasty (DMEK) and Descemet stripping automated endothelial keratoplasty (DSAEK) in the fellow eye of the same patients. Single-center, retrospective case series. Seventeen patients with bilateral Fuchs endothelial dystrophy who underwent
Saleh Sulaimana et al.
Environmental technology, 35(13-16), 1945-1955 (2014-06-25)
Stability and removal of dexamethasone sodium phosphate (DSP) from wastewater produced at Al-Quds University Campus were investigated. Kinetic studies in both pure water and wastewater coming from secondary treatment (activated sludge) demonstrated that the anti-inflammatory DSP underwent degradation to its

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service