Skip to Content
Merck
All Photos(1)

Key Documents

1019701

USP

Amiloride hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Amiloride hydrochloride dihydrate, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClN7O · HCl · 2H2O
CAS Number:
Molecular Weight:
302.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amiloride

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1nc(c(nc1N)N)C(=O)N\C(=N\[H])\N.Cl.O.O

InChI

1S/C6H8ClN7O.ClH.2H2O/c7-2-4(9)13-3(8)1(12-2)5(15)14-6(10)11;;;/h(H4,8,9,13)(H4,10,11,14,15);1H;2*1H2

InChI key

LTKVFMLMEYCWMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amiloride hydrochloride is a photosensitive yellow powder used as a mild diuretic drug.

Application

Amiloride hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amiloride Hydrochloride and Hydrochlorothiazide Tablets
  • Amiloride Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hector Rasgado-Flores et al.
American journal of physiology. Cell physiology, 305(11), C1114-C1122 (2013-08-30)
Hypertonic saline (HS) inhalation therapy benefits cystic fibrosis (CF) patients [Donaldson SH, Bennet WD, Zeman KL, Knowles MR, Tarran R, Boucher RC. N Engl J Med 354: 241-250, 2006; Elkins MR, Robinson M, Rose BR, Harbour C, Moriarty CP, Marks
Matthias Schoenberger et al.
Expert opinion on therapeutic patents, 23(10), 1383-1389 (2013-08-21)
Novel molecular platforms for epithelial sodium channel (ENaC) modulators are claimed in the following six patents: WO2012035158(A1); WO2009074575(A2); WO2011028740(A1); WO2009150137(A2); WO2011079087(A1); WO2008135557(A1). These ENaC inhibitors may be used in blocking transepithelial sodium and consequently water absorption across airway epithelia. This
Robert W Hunter et al.
American journal of physiology. Renal physiology, 306(4), F457-F467 (2014-01-10)
Na(+) reabsorption from the distal renal tubule involves electroneutral and electrogenic pathways, with the latter promoting K(+) excretion. The relative activities of these two pathways are tightly controlled, participating in the minute-to-minute regulation of systemic K(+) balance. The pathways are
Yusuke Ito et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 306(5), R315-R327 (2014-01-10)
Zebrafish Na(+)/H(+) exchanger 3b (zNHE3b) is highly expressed in the apical membrane of ionocytes where Na(+) is absorbed from ion-poor fresh water against a concentration gradient. Much in vivo data indicated that zNHE3b is involved in Na(+) absorption but not
Juraj Rievaj et al.
American journal of physiology. Gastrointestinal and liver physiology, 305(4), G303-G313 (2013-06-15)
Intestinal calcium (Ca²⁺) absorption occurs via paracellular and transcellular pathways. Although the transcellular route has been extensively studied, mechanisms mediating paracellular absorption are largely unexplored. Unlike passive diffusion, secondarily active paracellular Ca²⁺ uptake occurs against an electrochemical gradient with water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service