Skip to Content
Merck
All Photos(1)

Key Documents

129876

Sigma-Aldrich

Amiloride, Hydrochloride

An inhibitor of angiogenesis.

Synonym(s):

Amiloride, Hydrochloride, Acid-Sensing Ion Channel Modulator I, ASIC Channel Modulator I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClN7O · xHCl
CAS Number:
Molecular Weight:
229.63 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white to yellow

solubility

water: 2.5 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C6H8ClN7O.ClH/c7-2-4(9)13-3(8)1(12-2)5(15)14-6(10)11;/h(H4,8,9,13)(H4,10,11,14,15);1H

InChI key

ACHKKGDWZVCSNH-UHFFFAOYSA-N

General description

An inhibitor of angiogenesis. At low concentrations, acts as a potent and specific inhibitor of transmembrane Na+ entry and Na+/H+ ATPase. Inhibits smooth muscle responses to contractile stimuli. Blocks palytoxin-induced conductance in skeletal muscle. Reduces the electrical potential across tubular epithelium. At higher concentrations, blocks the Na+/H+ exchange pathway.
An inhibitor of angiogenesis. Inhibits capillary morphogenesis completely and reversibly at approximately 130 µM. At low concentrations, acts as a potent and specific inhibitor of transmembrane Na+ entry and Na+,K+-ATPase. Also acts as an inhibitor of urokinase-type plasminogen activator (uPA). Blocks smooth muscle responses to contractile stimuli and palytoxin-induced conductance in skeletal muscle. Reduces electrical potential across tubular epithelium. At higher concentrations, blocks the Na+/H+ exchange pathway.

Biochem/physiol Actions

Cell permeable: no
Primary Target
capillary morphogenesis
Product does not compete with ATP.
Reversible: yes
Target IC50: 130 µM inhibiting capillary morphogenesis completely and reversibly

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Knoll, A., et al. 1999. Microvasc. Res.58, 1.
Swiercz, R., et al. 1999. Oncol. Rep.6, 523.
Moran, A., et al. 1995. Am. J. Physiol. 268, C350.
Alliegro, M.C., et al. 1993. J. Exp. Zool. 267, 245.
Sugiyama, S., et al. 1988. J. Biol. Chem.263, 8215.
Yoshida, K., et al. 1988. Biochem. Biophys. Res. Commun.154, 101.
Sariban-Sotorabay, S., and Benos, D.J. 1986. Am. J. Physiol.250, C175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa M Reyes-Reyes et al.
Methods in molecular biology (Clifton, N.J.), 2036, 173-186 (2019-08-15)
Oligonucleotides can be designed or evolved to bind to specific DNA, RNA, protein, or small molecule targets and thereby alter the biological function of the target. The therapeutic potential of oligonucleotides targeted to intracellular molecules will depend largely on their
Dylan Adams et al.
PloS one, 17(5), e0266890-e0266890 (2022-05-04)
Sodium-hydrogen exchangers (NHEs) tightly regulate intracellular pH (pHi), proliferation, migration and cell volume. Heterogeneity exists between pulmonary endothelial cells derived from different vascular segments, yet the activity and isoform expression of NHEs between these vascular segments has not been fully
Ningyong Xu et al.
PloS one, 16(4), e0250095-e0250095 (2021-04-24)
Collective migration of endothelial cells is important for wound healing and angiogenesis. During such migration, each constituent endothelial cell coordinates its magnitude and direction of migration with its neighbors while retaining intercellular adhesion. Ensuring coordination and cohesion involves a variety
Juliette L Simonin et al.
Cells, 11(9) (2022-05-15)
Defective hydration of airway surface mucosa is associated with recurrent lung infection in cystic fibrosis (CF), a disease caused by CF transmembrane conductance regulator (CFTR) gene mutations. Whether the composition and/or presence of an airway surface liquid (ASL) is sufficient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service