Skip to Content
Merck
All Photos(1)

Documents

CRM51838

Supelco

2,4,5-T

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2,4,5-Trichlorophenoxyacetic acid, 2,4,5-T

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OCH2CO2H
CAS Number:
Molecular Weight:
255.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

154-158 °C (lit.)

format

neat

storage temp.

−20°C

SMILES string

OC(=O)COc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2H,3H2,(H,12,13)

InChI key

SMYMJHWAQXWPDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com2,4,5-T is a synthetic auxin that belongs to the family of chlorinated acid herbicides, used to defoliate broad-leafed plants.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, 2,4,5-T is not approved for use in the European Union. But a default maximum residue limit (MRL) of 0.01 mg/kg is applicable for the presence of 2,4,5-T (taken as a sum of 2,4,5-T, its salts, and esters) in products of plant origin, in accordance with Regulation (EC) No. 2015/868.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The 2,4,5-T CRM may also find use in the following:

  • Development of a multi-residue method for the determination of 24 plant growth regulators in 50 grape samples using a modified QuEChERS method and liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Development and validation of a method that included acidified QuEChERS extraction and LC-MS/MS determinationͰ based on fast switching between positive and negative electrospray ionization modes, to analyze 35 acid herbicides in food matrices
  • Application of a novel amino-modified covalent organic framework (NH2@COF) as a solid phase extraction (SPE) absorbent to extract carboxylic acid pesticides from environmental water samples
  • Determination of 12 phenoxy carboxylic acids in water samples by using a metal-organic framework (MIL-101) for dispersive solid phase extraction (DSPE) followed by ultra-high performance liquid chromatographic-tandem mass spectrometric (UHPLC-MS/MS) analysis
  • Analysis of 26 acid herbicides in 309 soil samples by dispersive solid-phase extraction and liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Investigating the use of magnetic polyethyleneimine-modified reduced graphene oxide (Fe3O4@PEI-RGO) as an adsorbent for the magnetic solid-phase extraction and HPLC determination of polar acidic herbicides in rice samples

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Li et al.
Analytica chimica acta, 949, 23-34 (2016-11-24)
A novel magnetic polyethyleneimine modified reduced graphene oxide (Fe3O4@PEI-RGO) had been fabricated based on a self-assemble approach between positive charged magnetic polyethyleneimine (Fe3O4@PEI) and negative charged GO sheets via electrostatic interaction followed by chemical reduction of GO to RGO. The
Chris Sack et al.
Journal of agricultural and food chemistry, 63(43), 9657-9665 (2015-10-17)
A method for the determination of 35 acid herbicides in food matrices was developed, validated, and implemented. It utilizes a modified QuEChERS extraction procedure coupled with quantitation by liquid chromatography tandem mass spectrometry (LC-MS/MS). The acid herbicides analyzed are all
Piotr Kaczyński et al.
Talanta, 152, 127-136 (2016-03-20)
This study determined twenty six the highly sensitive phenoxy, pyridines, aliphatic and aromatic acid compounds in soil with a liquid chromatography tandem mass spectrometry. The samples were prepared by modified quick, easy, cheap, effective, rugged and safe (QuEChERS) analytical procedure
Wen-Hua Ji et al.
Journal of chromatography. A, 1595, 11-18 (2019-03-06)
The amino-modified covalent organic framework (NH2@COF) was synthesized via the thiol-ene click reaction of vinyl covalent organic framework (COF) with 4-aminobenzenethiol. The introduction of amino groups can interact with the anionic headgroup of carboxylic acid pesticides. Moreover, ample hydrophilic surface
Zhongyin Ji et al.
Talanta, 191, 313-323 (2018-09-29)
A rapid and efficient method for extraction of 12 phenoxy carboxylic acids (PCAs) in environmental water samples was established based on metal-organic framework MIL-101 assisted dispersive solid phase extraction (DSPE). 12 PCAs were labeled by d0-10-methyl-2-(piperazin-1-ylsulfonyl)anthracen-9(10H)-one(d0-MASPz) and d3-10-methyl-2-(piperazin-1-ylsulfonyl)anthracen-9(10H)-one(d3-MASPz), allowing each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service