Skip to Content
Merck
All Photos(3)

Key Documents

135585

Sigma-Aldrich

4-Chlorobenzoic acid

99%

Synonym(s):

4-CBA, p-Chlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Beilstein:
907196
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

238-241 °C (lit.)

solubility

methanol: soluble 1%, clear, colorless to faintly yellow

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1ccc(Cl)cc1

InChI

1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)

InChI key

XRHGYUZYPHTUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chlorobenzoic acid is a degradation product of indomethacin. It is degraded by Acinetobacter sp. strain ST-1 and causes its dehalogenation to yield 4-hydroxybenzoic acid under both aerobic and anaerobic conditions.

Application

4-Chlorobenzoic acid can be used:
  • As a ligand to synthesize luminescent lanthanide complexes for bio-labeling or fiber communication applications.
  • To prepare organotin(IV) chlorobenzoates exhibiting anticorrosion properties.
  • As a ligand to synthesize di-n-butyl(4-chlorobenzoxy)(4-chlorobenzohydroxamato)tin(IV).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

460.4 °F - closed cup

Flash Point(C)

238 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Lindenauer et al.
Biomedical engineering, 11(9), 301-306 (1976-09-01)
Knitted Dacron cloth fabricated into tubes has gained widespread acceptance as an arterial prosthesis. Continued research and development in the field of vascular prostheses is required because existing materials are not ideal. This paper relates general developments in vascular prosthesis
Kikuo Kobayashi et al.
The Journal of general and applied microbiology, 43(2), 105-108 (1997-04-01)
Acinetobacter sp. strain ST-1, isolated from garden soil, can mineralize 4-chlorobenzoic acid (4-CBA). The bacterium degrades 4-CBA, starting with dehalogenation to yield 4-hydroxybenzoic acid (4-HBA) under both aerobic and anaerobic conditions, suggesting that the dehalogenating enzyme in the strain is
Ruiyang Xiao et al.
Environmental pollution (Barking, Essex : 1987), 257, 113498-113498 (2019-11-26)
Carbamazepine (CBZ), a widely detected pharmaceutical in wastewaters, cannot currently be treated by conventional activated sludge technologies, as it is highly resistant to biodegradation. In this study, the degradation kinetics and reaction mechanisms of CBZ by hydroxyl radical (OH) and
Q Q Cai et al.
Water research, 183, 116119-116119 (2020-07-15)
Fluidized bed reactor Fenton (FBR-Fenton) process was adopted for reverse osmosis concentrate (ROC) treatment with three types of carriers, including sand, zeolite and granular activated carbon (GAC). Adsorption studies demonstrated that GAC achieved the best adsorption performance (maximum COD removal
Aliyeh Hasanzadeh et al.
Scientific reports, 9(1), 13780-13780 (2019-09-26)
Nanocarbon materials are considered to be active for electrochemical oxygen reduction reaction (ORR) for hydrogen peroxide (H2O2) synthesis. In the present work, a new type of fullerene 60 (C60)-carbon nanotubes (CNTs) hybrid with covalently attached C60 onto outer surface of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service