Skip to Content
Merck
All Photos(1)

Key Documents

59343

Supelco

SUPELCOGEL Pb (9 µm) HPLC Columns

L × I.D. 30 cm × 7.8 mm

Synonym(s):

SUPELCOGEL 6% Cross Linked HPLC Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOGEL Pb, 6% Crosslinked HPLC Column, 9 μm particle size, L × I.D. 30 cm × 7.8 mm

Agency

suitable for USP L34

manufacturer/tradename

SUPELCOGEL

parameter

400 bar pressure (5801 psi)
85 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

sulfonated polystyrene/divinylbenzene, spherical particle platform

particle size

9 μm

separation technique

ligand exchange

Looking for similar products? Visit Product Comparison Guide

Application

SUPELCOGEL Pb, 6% Crosslinked HPLC Column has been used for the separation of:
  • D-mannose in human serum samples during its determination using liquid chromatography-tandem mass spectrometry (LC-MS/MS) equipped with electrospray ionization (ESI) source and selected reaction monitoring (SRM) detection.
  • Free sugars in switchgrass extractives during their quantification using high performance liquid chromatography equipped with refractive index (HPLC-RI) detector.
  • Total hemicellulose and cellulose sugars in sugarcane straw hydrolyzate during their determination using HPLC equipped with infra-red (IR) detector.

Legal Information

SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC

guard cartridge


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monitoring switchgrass composition to optimize harvesting periods for bioenergy and value-added products.
Lindsey K, et al.
Biomass and Bioenergy, 56, 29-37 (2013)
Sugars and char formation on subcritical water hydrolysis of sugarcane straw.
Lachos-Perez D, et al.
Bioresource Technology, 243, 1069-1077 (2017)
Lyndsey White et al.
Journal of pharmaceutical and biomedical analysis, 137, 54-59 (2017-01-17)
Several metabolites in human serum have been identified as potential cancer biomarkers for early detection. This study focuses on the LC-MS/MS method development and validation of d-mannose in human serum. Surrogate blank serum, coupled with stable isotope d-mannose-13C6, as internal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service