Skip to Content
Merck
All Photos(2)

Key Documents

53822-U

Supelco

Ascentis® Express 90 Å C18 (2.7 μm) HPLC Columns

L × I.D. 5 cm × 2.1 mm, HPLC Column

Synonym(s):

Core-shell (SPP) Fused Core C18 HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 2.1 mm

material

stainless steel column

Agency

suitable for USP L1

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

60 °C temp. range
600 bar max. pressure (9000 psi)

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

5 cm × 2.1 mm

surface area

135 m2/g

impurities

<5 ppm metals

matrix

Fused-Core particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2.7 μm

pore size

90 Å

operating pH

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Ascentis Express HPLC columns, through the use of Fused-Core® particle technology, can provide you with both the high speed and high efficiencies of sub-2 μm particles while maintaining lower backpressures. The combination of high efficiency and low backpressure benefits UPLC® (or other ultra high pressure system) users, as well as conventional HPLC users.
Visit the Ascentis Express home page for more information on this new column technology.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
UPLC is a registered trademark of Waters

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Foo Wong et al.
Electrophoresis (2018-02-08)
Metabolic profiling of Glycyrrhiza glabra using comprehensive two-dimensional liquid chromatography (LC × LC) coupled with photodiode array (PDA) and mass spectrometry (MS) detection is described. The separation was conducted under reversed-phase conditions, using a combination of first dimension (1 D) 150 mm microbore
Silvia Jakabová et al.
Journal of chromatography. A, 1232, 295-301 (2012-03-07)
Hyoscyamine (atropine) and scopolamine are the predominant tropane alkaloids in the Datura genus, occurring in all plant organs. The assessment of the alkaloid content of various plant parts is essential from the viewpoint of medical use, but also as a
Heena et al.
Journal of chromatographic science, 53(5), 800-806 (2014-10-18)
This study represents a new analytical high-performance liquid chromatography-fluorescence detector method for the determination of Al(III) as Al(III) complex with 8-hydroxyquinoline-5-sulfonic acid in a tap water sample and a coke sample. A micellar liquid chromatographic method is proposed for the
Petra Šilarová et al.
Food chemistry, 237, 471-480 (2017-08-03)
The degradation of catechins and other phenolics in green tea infusions were monitored using fast HPLC/MS separation. The final separation was performed within 2.5min using Ascentis Express C18 column (50mm×2.1mm i.d.) packed with 2μm porous shell particles. Degradation was studied
Babu Rao Chandu et al.
SpringerPlus, 2(1), 194-194 (2013-06-07)
A bioequivalence study was proved of generic Febuxostat 80 mg tablets (T) in healthy volunteers.For this purpose, Authors developed a simple, sensitive, selective, rapid, rugged and reproducible liquid chromatography-tandem mass spectrometry method for the quantification of Febuxostat (FB) in human

Articles

Improved LC/MS Detection of Estradiol using 4-(Dimethylamino) benzoyl chloride (DMABC) derivatization agent relative to dansyl chloride.

Accurate protein binding affinities were measured using bioSPME more rapidly than membrane dialysis (RED) techniques. Binding affinities correlated to the reference values.

Learn how filter selection and sample and mobile phase filtration impact HPLC back pressure and column lifetime.

Learn how filter selection and sample and mobile phase filtration impact HPLC back pressure and column lifetime.

See All

Protocols

We presents a robust method for analyzing 25-hydroxyvitamin D3 and 25-hydroxyvitamin D2 in serum using a multiplexed LC-MS-MS system.

Chromatograms

application for SPE, application for LC-MSapplication for HPLCapplication for HPLCapplication for HPLCShow More

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service