Merck
  • Development of a rapid and sensitive method for the determination of aluminum by reverse-phase high-performance liquid chromatography using a fluorescence detector.
Development of a rapid and sensitive method for the determination of aluminum by reverse-phase high-performance liquid chromatography using a fluorescence detector.
Journal of chromatographic science (2014-10-18)
Heena, Rajesh Kumar, Susheela Rani, Ashok Kumar Malik
ABSTRACT
This study represents a new analytical high-performance liquid chromatography-fluorescence detector method for the determination of Al(III) as Al(III) complex with 8-hydroxyquinoline-5-sulfonic acid in a tap water sample and a coke sample. A micellar liquid chromatographic method is proposed for the determination of aluminum metal in the presence of cetyltrimethylammonium bromide, a cationic surfactant (0.05 M) used for the solubilization of the aluminum complex. The influence of pH and ligand concentration on the formation of the complex was studied by adding a small amount of 0.1 M sodium hydroxide. The metal chelate was detected at λEx 410 nm and λEm 510 nm. This method eliminates the need for addition of reagent or organic modifier to the mobile phase. The complex was analyzed using an Ascentis Express C18 column and a mobile phase consisting of acetonitrile, methanol and water (55 : 30 : 15). Under the optimized conditions, the linear range was 1-200 µg L(-1) and the limit of detection was 0.05 µg L(-1). The method showed a good detector response over the range of interest and was successfully applied for the determination of trace Al(III) in canned coke and water samples containing excess of Mg(II), Ca(II) and other matrices.
Product Description
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 500 μm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
Sigma-Aldrich
Aluminum sulfate, meets USP testing specifications
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 10 mm
Supelco
Ascentis® Express C18, 2.7 μm HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express C18 PCP HPLC Columns, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
Sigma-Aldrich
Aluminum sulfate, 99.99% trace metals basis
Supelco
Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm