Skip to Content
Merck
All Photos(1)

Key Documents

47869

Supelco

Cyanocobalamin (B12)

analytical standard

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Beilstein:
4122889
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (decomposes before melting)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyanocobalamin (B12) is a water soluble vitamin, which belongs to the B12 class of vitamins, commonly called as cobalamins.(1) It is mainly found in animal products such as dairy products, meat and also eggs.[1] It can find applications in the preparation of multivitamin supplements and fortified food.[2] It works as a coenzyme to convert L-methylmalonyl CoA to succinyl CoA, which is catalyzed by isomerase.[2]

Application

Cyanocobalamin (B12) may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations and food products by various chromatography techniques.[3][4][5][6][7][8]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shreemathy V and Dandekar PS
Nutrition and Biochemistry for Nurses, (2) (2015)
Cyanocobalamin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 20(3), 2178-2179 (2014)
Determination of cyanocobalamin by isotope dilution LC-MS/MS
D-Ulivo L, et al.
Analytica Chimica Acta (2017)
A rapid determination of the relative purity of vitamin B12 (Cyanocobalamin) in pharmaceutical products
Bruening CF, et al.
Journal of the American Pharmaceutical Association, 47(1), 15-20 (1958)
The assay of cyanocobalamin in pharmaceutical preparations by solid-phase extraction and HPLC
Jansen CC and De Kleijn JP
Journal of Chromatographic Science, 28(1), 42-45 (1990)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service