Skip to Content
Merck
All Photos(5)

Documents

C0884

Sigma-Aldrich

Coenzyme B12

≥97.0%

Synonym(s):

5′-Deoxyadenosylcobalamine, Adenosyl cobalamine, Cobamamide, DMBC coenzyme

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H100CoN18O17P
CAS Number:
Molecular Weight:
1579.58
Beilstein:
4122932
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

Quality Level

Assay

≥97.0%

form

powder and chunks

technique(s)

HPLC: suitable

color

red to very dark red
very dark red-brown

storage temp.

−20°C

SMILES string

CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N([Co+]C[C@H]3O[C@@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)C1=C(C)C6=NC(=CC7=NC(=C(C)C8=NC2(C)C(C)(CC(N)=O)C8CCC(N)=O)C(C)(CC(N)=O)C7CCC(N)=O)C(C)(C)C6CCC(N)=O)OP([O-])(=O)O[C@H]9[C@@H](O)[C@H](O[C@@H]9CO)n%10cnc%11cc(C)c(C)cc%10%11

InChI

1S/C62H90N13O14P.C10H12N5O3.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-4-6(16)7(17)10(18-4)15-3-14-5-8(11)12-2-13-9(5)15;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);2-4,6-7,10,16-17H,1H2,(H2,11,12,13);/q;;+2/p-2/t31?,34?,35?,36?,37?,41-,52-,53-,56?,57+,59?,60?,61?,62?;4-,6-,7-,10+;/m11./s1

InChI key

ZIHHMGTYZOSFRC-PMDPKNCCSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin B12 cobalamin refers to a group of chemically-related cobalt containing molecules. The physiologically active forms of vitamin B12 include methylcobalamin and adenosylcobalamin, whereas hydroxocobalamin (vitamin B12a, OHCbl) and cyanocobalamin (CNCbl) are storage and delivery forms.

Application

Coenzyme B12 has been used as a supplement for culturing plasmid variants in in vivo assay of yvrC-lacZ fusions.

Biochem/physiol Actions

Vitamin B12 cobalamin is involved in DNA synthesis and fatty acid synthesis. It also plays a vital role as a coenzyme in the conversion of mitochondrial methylmalonyl co-enzyme A to succinyl co-enzyme A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis Bucher et al.
Journal of the American Chemical Society, 134(3), 1591-1599 (2012-01-11)
Vitamin B(12) and its biologically active counterparts possess the only examples of carbon-cobalt bonds in living systems. The role of such motifs as radical reservoirs has potential application in future catalytic and electronic nanodevices. To fully understand radical generation in
J R Roth et al.
Annual review of microbiology, 50, 137-181 (1996-01-01)
This review examines deoxyadenosylcobalamin (Ado-B12) biosynthesis, transport, use, and uneven distribution among living forms. We describe how genetic analysis of enteric bacteria has contributed to these issues. Two pathways for corrin ring formation have been found-an aerobic pathway (in P.
Peter Friedrich et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(50), 16114-16122 (2012-10-20)
Coenzyme B(12) can assist radical enzymes that accomplish the vicinal interchange of a hydrogen atom with a functional group. It has been proposed that the Co-C bond homolysis of coenzyme B(12) to cob(II)alamin and the 5'-deoxyadenosyl radical is aided by hydrogen bonding
Alla Peselis et al.
Nature structural & molecular biology, 19(11), 1182-1184 (2012-10-16)
Coenzyme B(12) has a key role in various enzymatic reactions and controls expression of bacterial genes through riboswitches. Here we report the crystal structure of the Symbiobacterium thermophilum B(12) riboswitch bound to its ligand adenosylcobalamin. The riboswitch forms a unique
Jiayun Pang et al.
Journal of the American Chemical Society, 134(4), 2367-2377 (2012-01-14)
We present here an energetic and atomistic description of how D-ornithine 4,5-aminomutase (OAM), an adenosylcobalamin (AdoCbl; coenzyme B(12))-dependent isomerase, employs a large-scale protein domain conformational change to orchestrate the homolytic rupture of the Co-C bond. Our results suggest that in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service