Skip to Content
Merck
All Photos(1)

Key Documents

40669

Supelco

Cumene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C9H12/c1-8(2)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

RWGFKTVRMDUZSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cumene is chemical, which can be released into the environment due to production and processing , from petroleum refining and the combustion and evaporation of petroleum products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Environmental Fate and Exposure Data For Organic Chemicals, 5 (1997)
Johanna Bråred Christensson et al.
Contact dermatitis, 55(4), 230-237 (2006-09-09)
Concomitant positive reactions to colophonium, oxidized limonene, and/or oxidized linalool are recorded in patch test studies. The main allergens in these patch test mixtures are hydroperoxides, which form antigens by a radical pathway. Theoretically, concomitant reactions can be explained not
National Toxicology Program technical report series, (542)(542), 1-200 (2009-04-03)
Cumene occurs naturally in petroleum and is used as a solvent, in gasoline and diesel fuels, and as the principal chemical in the production of phenol and acetone. We studied cumene to determine if it caused cancer in rats or
V I Lushchak et al.
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 78(6), 113-120 (2007-05-15)
Two methods of the determination of lipid peroxidation products have been compared which are based on Fe(II) oxidation by them at acid pH values in the presence of xylenol orange which binds Fe(III) have been compared. The first method uses
C Holvoet et al.
Die Pharmazie, 62(2), 126-132 (2007-03-08)
Paclitaxel, an antitumoral drug, is poorly soluble in aqueous media. Therefore, in a commercialised formulation (Taxol), paclitaxel (30 mg active compound) is dissolved in polyethoxylated castor oil (Cremophor EL) and ethanol. After dilution of Taxol in aqueous media paclitaxel tends

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service