Skip to Content
Merck
All Photos(1)

Key Documents

502111

Supelco

EPA 502/524 Volatiles Organic Calibration Mix (without gases)

2000 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
Pricing and availability is not currently available.

grade

analytical standard

Agency

EPA 502.1,524.1,524.2

description

Separate Source
Without gases

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Application

EPA 502/524 volatiles organic calibration mix has been used as a standard in the analysis of contaminants like volatile organic compounds in food product samples by using gas chromatography coupled with mass spectrometry (GC-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene
    Bromobenzene
    Bromochloromethane
    Bromodichloromethane
    Bromoform
    Butylbenzene
    sec-Butylbenzene
    tert-Butylbenzene
    Carbon tetrachloride
    Chlorobenzene
    Chloroform
    2-Chlorotoluene
    4-Chlorotoluene
    Dibromochloromethane
    1,2-Dibromo-3-chloropropane
    1,2-Dibromoethane
    Dibromomethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    1,3-Dichloropropane
    2,2-Dichloropropane
    1,1-Dichloro-1-propene
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Ethylbenzene
    Hexachloro-1,3-butadiene
    Cumene
    p-Cymene
    Naphthalene
    Propylbenzene
    Styrene
    1,1,1,2-Tetrachloroethane
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,3-Trichlorobenzene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    1,2,3-Trichloropropane
    1,2,4-Trimethylbenzene
    Mesitylene
    m-Xylene
    p-Xylene
    o-Xylene
    See All (54)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system, Liver,Kidney

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Occurrence of volatile organic compounds in foods from the Belgian market and dietary exposure assessment
Vinci MR, et al.
Food Control, 52, 1-8 (2015)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service