Skip to Content
Merck
All Photos(1)

Key Documents

40346

Supelco

Dichlorodifluoromethane solution

5000 μg/mL in methanol, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl2F2
CAS Number:
Molecular Weight:
120.91
UNSPSC Code:
41116105

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CCl2F2/c2-1(3,4)5

InChI key

PXBRQCKWGAHEHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Ozone 1 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Hsiang Lo et al.
Environmental health perspectives, 114(11), 1713-1717 (2006-11-17)
Although there are ample data on the respiratory effects of exposure to fire extinguisher gas, the potential hematologic effects have not been fully documented. We conducted this study to determine the possible etiologic agent(s) for a decrease in red blood
David M Wilmouth et al.
The journal of physical chemistry. A, 113(51), 14099-14108 (2009-10-27)
Recent laboratory measurements of the absorption cross sections of the ClO dimer, ClOOCl, have called into question the validity of the mechanism that describes the catalytic removal of ozone by chlorine. Here we describe direct measurements of the rate-determining step
Yukinori Maruo et al.
Dental materials journal, 24(3), 311-314 (2005-11-11)
The purpose of this study was to examine if a modified direct relining method could result in excellent denture adaptation compared to a conventional method. Direct relining was done using heat-polymerized acrylic resin base on a maxillary cast with two
Tian-yang Xiu et al.
Huan jing ke xue= Huanjing kexue, 26(1), 1-6 (2005-04-30)
In recent years, concentration of CFC in the atmosphere has undergone rapid changes especially in association with human activities. For the influence of CFC on climate changes and environment changes, the concentration of atmospheric CFC-11 in Beijing is continuously observed
S-K Wang et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(43), 435011-435011 (2011-10-15)
Continuous-time core-level photon-stimulated desorption (PSD) spectroscopy was used to investigate the monochromatic soft x-ray photoreactions of CF(2)Cl(2) adsorbed on Si(111)-7 × 7 near the F(1s) edge (681-704 eV). Sequential F(+) PSD spectra were observed as a function of photon exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service