Skip to Content
Merck
All Photos(1)

Key Documents

40156

Supelco

Pentachloronitrobenzene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Agency

EPA 508,508A,8080,8081,8082,508.1

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9

InChI key

LKPLKUMXSAEKID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup - Solvent

Flash Point(C)

9.7 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Alborch et al.
Letters in applied microbiology, 50(3), 270-275 (2010-01-15)
To assess differences between two recommended selective culture media, Nash and Snyder medium (NS) and malachite green agar 2.5 (MGA 2.5), for the detection of Fusarium infection in conventional and transgenic maize kernels. In total, 10 800 kernels from commercial
Wang Zhang et al.
Journal of hazardous materials, 198, 275-281 (2011-11-15)
Pentachloronitrobenzene (PCNB) has been identified as a potential persistent organic pollutant, and received concerns from many countries for regulation due to its impurities such as dioxins and furans. Considering the need for its disposal, the mechanochemical destruction approach was investigated
Weiguo Zhang et al.
Rapid communications in mass spectrometry : RCM, 20(10), 1563-1568 (2006-04-22)
The effects of overlapping levels and concentration ratios of overlapping components, and of scan rates of the mass spectrometer, on the capability of the automated mass spectral deconvolution and identification system (AMDIS) in pesticide residue analysis were studied. To investigate
Kazuhiro Takagi et al.
Applied and environmental microbiology, 75(13), 4452-4458 (2009-05-12)
A novel aerobic pentachloronitrobenzene-degrading bacterium, Nocardioides sp. strain PD653, was isolated from an enrichment culture in a soil-charcoal perfusion system. The bacterium also degraded hexachlorobenzene, a highly recalcitrant environmental pollutant, accompanying the generation of chloride ions. Liberation of (14)CO(2) from
C Michielsen et al.
Archives of toxicology, 74(10), 609-617 (2001-02-24)
Involvement of the mercapturic acid pathway in the induction of splenomegaly and skin and lung pathology by hexachlorobenzene (HCB) in the rat was investigated by seeking to determine whether pentachloronitrobenzene (PCNB) has the same inflammatory effects as HCB, since both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service