Skip to Content
Merck
All Photos(1)

Key Documents

20350

Supelco

ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg

RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea

Synonym(s):

ORBO Supelpak 20 series Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900
Pricing and availability is not currently available.

material

RC,GFF,F,F (specially cleaned) separators

Quality Level

Agency

NIOSH 5601
OSHA 62,63,67,70,74

product line

Amberlite
ORBO

composition

Bed A, 270 mg
Bed B, 140 mg

packaging

pkg of 10 ea

manufacturer/tradename

ORBO 49P

technique(s)

active air sampling: suitable

O.D. × L

13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube

matrix

XAD-2 (specially cleaned)

particle size

20-40 mesh

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

The OSHA Versatile Sampler (OVS) was primarily designed for collection of Organophosphorus pesticides but is suitable for other applications. This specially constructed open glass tube is 75 mm long with a 13-mm outer diameter (O.D.) on the inlet end tapered to 8 mm (O.D.) on the outlet end. The design features a glass filter to trap the aerosols and a two-section sorbent portion to adsorb the vapors. The tube is packed with specially cleaned XAD-2 material and is QA tested to ensure low background interferences. The product is capped at the inlet and outlet to minimize environmental exposure. Meets OSHA and NIOSH method design specifications.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
ORBO is a trademark of Sigma-Aldrich Co. LLC
Supelpak is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Rochat et al.
Journal of chemical ecology, 17(11), 2127-2141 (1991-11-01)
Male American palm weevils (APWs),Rhynchophorus palmarum (L.) produced two sex-specific compounds, which were disclosed by volatile collections on Supelpak-2 and gas chromatography. One was a minor compound, not always detected. The major male-produced volatile was identified as (2E)-6-methyl-2-hepten-4-ol through coupled
F J Egea González et al.
Journal of AOAC International, 80(5), 1091-1097 (1997-11-05)
A method to sample and analyze chlorothalonil and dichlofluanid in greenhouse air was evaluated. Analysis was performed by gas chromatography with electron capture detection and gas chromatography-mass spectrometry. Solid sorbents such as Chromosorb 102, Porapak R, Supelpak-2, Amberlite XAD-2, Amberlite
Anne Ørbo et al.
The Journal of steroid biochemistry and molecular biology, 113(1-2), 139-149 (2009-01-14)
Patients with endometrial hyperplasia representing preliminary stages of endometrial cancer have shown to respond to therapy in 100% of the cases when treated with levonorgestrel-impregnated intrauterine device. Anti-proliferative effect has also been reported after application of an anti-progestin impregnated intrauterine
H Stalsberg et al.
Virchows Archiv. A, Pathological anatomy and histopathology, 414(5), 415-422 (1989-01-01)
The autopsies of 12 victims from two snow avalanches in North-Norway are reported. Supportive evidence from non-autopsied and surviving victims is included. Consistent autopsy findings were prominent lung oedema, moderate cerebral oedema, extreme contraction of the left ventricle, petechiae in
Anne Beate Vereide et al.
Gynecologic oncology, 97(3), 740-750 (2005-05-12)
The aim of the study was to investigate apoptosis as a growth regulatory mechanism of gestagen in endometrial precancers and to compare differences in the apoptotic cascade after high and low dose gestagen regimens. Pre- and post-treatment paraffin-embedded endometrial hyperplasia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service