Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1344

Sigma-Aldrich

Anti-Tec Antibody, clone 19M23, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Tyrosine-protein kinase Tec

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

19M23, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 73.58 kDa
observed mol wt ~65 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TEC(7006)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 19M23 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Tyrosine-protein kinase Tec. It targets an epitope within 19 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human Tyrosine protein kinase Tec.

Application

Affinity Binding Assay: A representative lot of this antibody bound Tec peptide with a KD of 1.2 x 10-5 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Tec in K562 cells.

ELISA Analysis: A representative lot specifically detected Tec peptide in ELISA application, but not peptides from five other proteins.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Anti-Tec, clone 19M23 ZooMAb, Cat. No. ZRB1344, is a recombinant Rabbit monoclonal antibody that specifically targets human Tyrosine Protein kinase Tec and has been tested for use in Affinity Binding Assay , ELISA, Immunocytochemistry, and Western Blotting.

Target description

Tyrosine-protein kinase Tec (UniProt: P42680; also known as EC:2.7.10.2, Tec) is encoded by the TEC (also known as PSCTK4) gene (Gene ID: 7006) in human. The Tec family of protein kinases consists of five members, Tec, Btk, Itk/Emt/Tsk, Bmx and Txk/Rlk. They are abundantly expressed in hematopoietic cells and participate in the growth and differentiation of blood cells. Tec is a peripheral membrane protein that serves as a non-receptor tyrosine kinase that contributes to signaling from many receptors and participates as a signal transducer in multiple downstream pathways, including regulation of the actin cytoskeleton. It is activated by tyrosine phosphorylation by a wide range of cytokine stimulations. It regulates the development, function and differentiation of conventional T-cells and nonconventional NKT-cells and is required for TCR-dependent IL2 gene induction. Following B-cell or T-cell receptors activation by antigen, it is shown to translocate to the plasma membrane through its PH domain (aa 4-111). Tec is reported to serve as a central regulator of the noncanonical caspase-8 inflammasome that is involve in the assembly and activation of the caspase-8 inflammasome in response to fungal pathogens but is not involved in anti-bacterial immune responses. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Mano, H., et al. (1999). Cytokines Growth Factor Rev. 10(3-4); 267-280; Zwolanek, F., et al. (2014). PLoS Pathog. 10(12); e1004525.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service