Key Documents
Z768650
Nalgene® square graduated media bottle
with cap, PET, sterile, capacity 650 mL
Synonym(s):
media storage bottle, square plastic bottle
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
capacity:
650 mL
material:
PET
square bottle
sterility:
sterile
feature:
graduations
with cap
packaging:
pack of 48 ea (2 x pack of 24)
material
PET
square bottle
Joint
Joints threaded neck (38-430)
sterility
sterile
feature
graduations
with cap
packaging
pack of 48 ea (2 x pack of 24)
manufacturer/tradename
Nalgene® 342040-0650 (USA)
Nalgene® 842040-0650
capacity
650 mL
Features and Benefits
  • Closure made up of high-density polyethylene (HDPE)
  • Transparent and leakproof
  • Sterilized by gamma radiation, Sterility Assurance Level (SAL) is 10-6
  • Decreased permeability to CO2 and O2
  • Non-cytotoxic and non-pyrogenic
  • Bottle and closure are non-animal derived
  • Graduated and neck ring for use with tamper-evident shrink sleeves
  • Excellent gas barrier properties for easy shipping and storage of buffers, liquid media, and sera
  • Light-weight, shatter-resistant, and space-saving square shape
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)