Skip to Content
Merck
All Photos(1)

Key Documents

Z768669

Nalgene® square graduated media bottle

with cap, PET, sterile, capacity 1000 mL

Synonym(s):

media storage bottle, square plastic bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17
capacity:
1000 mL
material:
PET
square bottle
sterility:
sterile
feature:
graduations
with cap
packaging:
pack of 24 ea (2 x pack of 12)

material

PET
square bottle

Joint

Joints threaded neck (38-430)

sterility

sterile

feature

graduations
with cap

packaging

pack of 24 ea (2 x pack of 12)

manufacturer/tradename

Nalgene® 342040-1000 (USA)
Nalgene® 842040-1000 (European)

capacity

1000 mL

Looking for similar products? Visit Product Comparison Guide

Application

Nalgene® square graduated media bottle is ideal for storing and shipping buffers, serums, and liquid media.

Features and Benefits

  • The bottle is made of polyethylene terephthalate and the closure is made up of natural high-density polyethylene (HDPE)
  • Graduated and the neck ring for use with tamper evident shrink sleeves
  • Sterilized by gamma radiation
  • Non-cytotoxic, non-pyrogenic and leakproof
  • Transparent and reduced permeability to carbon-di-oxide and oxygen
  • Bottle and closure are non-animal derived
  • Space-saving square designed, light-weight and non-breakable

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service