Skip to Content
Merck
All Photos(3)

Key Documents

Z2625

Sigma-Aldrich

Zomepirac sodium salt

Synonym(s):

5-(p-Chlorobenzoyl)-1,4-dimethylpyrrole-2-acetic acid sodium-potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H13ClNO3Na
CAS Number:
Molecular Weight:
313.71
EC Number:
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Quality Level

SMILES string

Cc1cc(CC(=O)O[Na])n(C)c1C(=O)c2ccc(Cl)cc2

InChI

1S/C15H14ClNO3.Na/c1-9-7-12(8-13(18)19)17(2)14(9)15(20)10-3-5-11(16)6-4-10;/h3-7H,8H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

SEEXPXUCHVGZGU-UHFFFAOYSA-M

Application

An NSAID. Circumvents MRP-mediated multidrug resistance. Significantly increases the cytotoxicity of the anthracyclines (doxorubicin, daunorubicin and epirubicin), as well as teniposide, VP-16 and vincristine.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Bailey et al.
Journal of pharmacological and toxicological methods, 41(1), 27-32 (1999-10-03)
The covalent binding of drugs or their metabolites to proteins is of increasing interest in the investigation of the toxicity of these compounds. Recent attention on biological consequences of protein adduct formation with carboxylate drugs, derived via their reactive acyl
Jørgen Olsen et al.
Chemical research in toxicology, 18(11), 1729-1736 (2005-11-23)
Zomepirac [ZP, 5-(chlorobenzoyl)-1,4-dimethylpyrrole-2-acetic acid] was withdrawn from the market because of unpredictable allergic reactions that may have been caused by ZP-protein adducts formed by reaction of the reactive acyl glucuronide of ZP (ZP-O-G) with endogenous proteins. To test the hypothesis
J Abraham
Social science & medicine (1982), 46(1), 39-51 (1998-02-17)
This article systematically examines government regulation of medicines in the U.K. and the U.S. with specific reference to carcinogenic risk assessment. By taking four non-steroidal anti-inflammatory drugs (NSAIDs) as case studies, it is argued that there have been inconsistencies between
M J Bailey et al.
Chemical research in toxicology, 9(3), 659-666 (1996-04-01)
Carboxylate drugs usually form acyl glucuronide conjugates as major metabolites. These electrophilic metabolites are reactive, capable of undergoing hydrolysis, rearrangement, and covalent binding reactions to proteins. The last-mentioned property has the potential to initiate immune and other toxic responses in
G R Cannell et al.
Life sciences, 70(1), 37-48 (2002-01-05)
Many nonsteroidal anti-inflammatory drugs (NSAIDs) which have antiproliferative activity in colon cancer cells are carboxylate compounds forming acyl glucuronide metabolites. Acyl glucuronides are potentially reactive, able to hydrolyse, rearrange into isomers, and covalently modify proteins under physiological conditions. This study

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service