Skip to Content
Merck
All Photos(4)

Documents

M7279

Sigma-Aldrich

N,N′-Methylenebisacrylamide

powder, for molecular biology, suitable for electrophoresis, ≥99.5%

Synonym(s):

BIS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
Beilstein:
1706297
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

grade

for molecular biology

Quality Level

Assay

≥99.5%

form

powder

technique(s)

electrophoresis: suitable

impurities

≤0.05% Free carboxylic acid (T)
≤50 mg/kg Total sulfur (as SO4 ) (ICP)

conductivity

≤10 μmho (2% solution)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤10 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cr: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤5 ppm
K: ≤50 ppm
Mg: ≤5 ppm
Mn: ≤5 ppm
Na: ≤50 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Zn: ≤5 ppm

absorption

≤0.2 at 290 at 1%

foreign activity

DNase and RNase, none detected

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Methylenebisacrylamide is commonly called as bis. It is used as a cross-linking agent in the preparation of polyacrylamide gels consisting of chains of acrylamide monomers. The concentration of N,N′-methylenebisacrylamide and acrylamide determines the pore size of the gel. The higher concentration of bis forms rigid gels with smaller pores due to more cross-linking leading to polymerization and vice versa.

Application

N,N′-Methylenebisacrylamide has been used:
  • as a crosslinker in the monomer solution to prepare N,N-dimethyl acrylamide (DMAAm)-based cryogel and acrylamide (AAm)-based cryogel matrix
  • as a crosslinker to prepare tough hydrogel network
  • in the preparation of acrylamide-bisacrylamide stock solution for the preparation of polyacrylamide gel

To be mixed with acrylamide for making polyacrylamide gels for use in protein and nucleic acid electrophoresis.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrastretchable Conductor Fabricated on Skin-Like Hydrogel-Elastomer Hybrid Substrates for Skin Electronics
Kim SH, et al.
Advanced Materials, 30(26), 1800109-1800109 (2018)
Depletion of hemoglobin and carbonic anhydrase from erythrocyte cytosolic samples by preparative clear native electrophoresis
Damici GM, et al.
Nature Protocols, 7(1), 36-36 (2012)
Fundamentals and Techniques of Biophysics and Molecular Biology (2018)
Ashok Kumar et al.
Nature protocols, 5(11), 1737-1747 (2010-10-30)
In cell affinity chromatography, type-specific cell separation is based on the interaction between cell-surface receptors and an immobilized ligand on a stationary matrix. This protocol describes the preparation of monolithic polyacrylamide and polydimethylacrylamide cryogel affinity matrices that can be used
Ghazaleh Haghiashtiani et al.
Science advances, 6(35), eabb4641-eabb4641 (2020-09-15)
Minimally invasive surgeries have numerous advantages, yet complications may arise from limited knowledge about the anatomical site targeted for the delivery of therapy. Transcatheter aortic valve replacement (TAVR) is a minimally invasive procedure for treating aortic stenosis. Here, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service