Skip to Content
Merck
All Photos(2)

Key Documents

Y1000

Sigma-Aldrich

Select Yeast Extract

powder, BioReagent, suitable for cell culture, suitable for insect cell culture

Synonym(s):

Cell culture tested Yeast Extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.75

Agency

suitable for SM 5210

product line

BioReagent

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

solubility

H2O: 10% (remains clear after heating to 40°C.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For general bacteriological use with a variety of microorganisms.
Select Yeast Extract has been used:
  • as a component of nutritive medium to culture the parasite from the egg to the third infective larval stage
  • in yeast extract peptone dextrose (YPD)-medium/YPD agar and buffered complex glycerol (BMGY/BMMY) medium to culture Pichia pastoris transformants
  • to prepare nutrient medium and growth medium for the larval development assay (LDA) using gastrointestinal nematode parasite eggs

Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Spray dried, autolyzed yeast extract.
Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tamim Al-Jubair et al.
STAR protocols, 3(2), 101298-101298 (2022-04-26)
Aquaporins (AQPs) are membrane-bound water channels that play crucial roles in maintaining the water homeostasis of the human body. Here, we present a protocol for high-yield recombinant expression of human AQPs in the methylotropic yeast Pichia pastoris and subsequent AQP
Abdul-Lateef Molan et al.
Folia parasitologica, 57(1), 62-68 (2010-05-11)
The effects of condensed tannins (CTs) extracted from five species of plants on egg hatching and larval development of Teladorsagia circumcincta (Stadelmann, 1894) (syn. Ostertagia circumcincta) were evaluated using in vitro bioassays. The extracts of CTs were obtained from Lotus
O Tibe et al.
Veterinary parasitology, 197(1-2), 160-167 (2013-08-10)
The effect of condensed tannins (CT) extracted from forage plants from Botswana on the free-living stages of a number of species of gastrointestinal nematode parasites derived from infected sheep were investigated using in vitro assays. Fresh samples of five different
Marianne Cousin et al.
PloS one, 8(5), e65693-e65693 (2013-06-01)
The effects of the herbicide Paraquat were investigated in honey bee larvae with attention focused on oenocytes. Honey bee larvae were exposed to Paraquat at different concentrations in the food: 0, 0.001, 0.01, 0.1 and 1 µg/kg. In controls, between
Louis Philip Benoit Bouvrette et al.
RNA (New York, N.Y.), 24(1), 98-113 (2017-10-29)
Cells are highly asymmetrical, a feature that relies on the sorting of molecular constituents, including proteins, lipids, and nucleic acids, to distinct subcellular locales. The localization of RNA molecules is an important layer of gene regulation required to modulate localized

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service