Skip to Content
Merck
All Photos(2)

Key Documents

Y0875

Sigma-Aldrich

Select Yeast Extract

for use in microbial growth medium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

grade

for molecular biology

sterility

non-sterile

form

powder

technique(s)

microbiological culture: suitable

pH

6.5-7.5(10% solution)

solubility

H2O: 100 mg/mL, clear to slightly hazy, yellow to brownish-yellow

application(s)

microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Yeast extract is the water-soluble portion of autolyzed yeast, often used in culture media. Yeast extract contains a mixture of amino acids, peptides, water soluble vitamins and carbohydrates.
Yeast extract is used in all solid media, on which stock cultures are maintained, due to its ability in promoting growth of most bacterial contaminants, thereby making them visible to identify. Yeast extract does not enhance the growth of wild-type cells but essential for auxotrophic mutants.

Application

Select Yeast Extract has been used:
  • as a supplement in the minimal sporulating medium (MSL) for the secretory production of human tumor necrosis factor-α (TNFα) using the carboxypeptidase Y (Cpy) signal peptide.
  • in the isolation of halophilic archaea.
  • as a component of microbial growth medium for cultivation of marine bacterial strain Zobellia uliginosa.

    Suitable for use as nutritional source in microbial media.
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.

Other Notes

Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterization and antimicrobial potential of extremely halophilic archaea isolated from hypersaline environments of the Algerian Sahara
Quadri I, et al.
Microbiological research, 186, 119-131 (2016)
Chlamydomonas in the Laboratory
The Chlamydomonas Sourcebook (2009)
Fermentation optimization, purification and biochemical characterization of a porphyran degrading enzyme with funoran side-activity from Zobellia uliginosa
Howlader M, et al.
Biocatalysis and agricultural biotechnology, 43.0, 102394-102394 (2022)
L Wu et al.
Journal of applied microbiology, 97(1), 93-103 (2004-06-10)
Isolation, identification and characterization of a highly efficient isomaltulose producer. After an enrichment procedure for bacteria likely to metabolize isomaltulose in sucrose-rich environments, 578 isolates were screened for efficient isomaltulose biosynthesis using an aniline/diphenylamine assay and capillary electrophoresis. An isolate
Comparison of different signal peptides for secretion of heterologous proteins in fission yeast
Kjaerulff S and Jensen MR
Biochemical and Biophysical Research Communications, 336(3), 974-982 (2005)

Protocols

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service