Skip to Content
Merck
All Photos(1)

Key Documents

XNAT2R

Sigma-Aldrich

Extract-N-Amp Tissue PCR Kit

sufficient for 1000 extractions, sufficient for 1000 amplifications

Synonym(s):

Tissue direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 1000 amplifications
sufficient for 1000 extractions
sufficient for 1000 reactions

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Tissue PCR Kits provide all the reagents necessary to rapidly extract DNA from a wide variety of cells and tissues and amplify targets of interest by direct PCR (polymerase chain reaction) . A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR ReadyMix for amplification directly from the extract.

Application

Extract-N-Amp tissue PCR kit has been used for DNA extraction and polymerase chain reaction (PCR).

Features and Benefits

  • Fast – rapid extraction of genomic DNA for PCR in 15 minutes
  • Convenient – no long enzymatic digestions or column purification required
  • Practical – perfect for quick genomic DNA isolation for genotyping
  • Flexible – protocols available for mouse-tails, hair, animal tissue, saliva, and buccal swabs
  • Specific – Hot start antibody for highly specific PCR amplification of genomic DNA
  • Extract stable at 4 oC for at least 6 months

Components

Kits contain Tissue Preparation Solution, Extraction Solution, and Extract-N-Amp PCR Reaction Mix.

Principle

The kit contains validated protocols to extract and amplify genomic DNA from mouse-tails, hair, animal tissue, saliva, and buccal swabs. In a typical procedure, genomic DNA is extracted from a sample that has been incubated in the tissue preparation solution and extraction solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and Extract-N-Amp PCR ReadyMix, included in the kit.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.
This kit includes a clear solution of PCR readymix. To order the REDExtract-N-Amp Tissue PCR Kits with the inert tracking and loading dye in the PCR readymix, please order XNAT-1000RXN"

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Boi et al.
Virology, 499, 136-143 (2016-09-23)
We have demonstrated in a mouse model that infection with a retrovirus can lead not only to the generation of recombinants between exogenous and endogenous gammaretrovirus, but also to the mobilization of endogenous proviruses by pseudotyping entire polytropic proviral transcripts
Cindy Y Kok et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 21(10), 1823-1831 (2013-07-03)
Viral vectors based on adeno-associated virus (AAV) are showing exciting promise in gene therapy trials targeting the adult liver. A major challenge in extending this promise to the pediatric liver is the loss of episomal vector genomes that accompanies hepatocellular
Sergio Y Alcoser et al.
BMC biotechnology, 11, 124-124 (2011-12-20)
Xenograft samples used to test anti-cancer drug efficacies and toxicities in vivo contain an unknown mix of mouse and human cells. Evaluation of drug activity can be confounded by samples containing large amounts of contaminating mouse tissue. We have developed
Xiaodong Sun et al.
Cell cycle (Georgetown, Tex.), 12(7), 1133-1141 (2013-03-09)
Parkin, an E3 ubiquitin ligase well known for its role in the pathogenesis of juvenile Parkinson disease, has been considered as a candidate tumor suppressor in certain types of cancer. It remains unknown whether parkin is involved in the development
Erin C Zook et al.
Blood, 118(22), 5723-5731 (2011-09-13)
The forkhead box n1 (Foxn1) transcription factor is essential for thymic organogenesis during embryonic development; however, a functional role of Foxn1 in the postnatal thymus is less well understood. We developed Foxn1 transgenic mice (Foxn1Tg), in which overexpression of Foxn1

Articles

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

See All

Protocols

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service