Skip to Content
Merck
All Photos(3)

Key Documents

XNAT2

Sigma-Aldrich

Extract-N-Amp Tissue PCR Kit

sufficient for 100 extractions, sufficient for 100 amplifications

Synonym(s):

Tissue direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 100 amplifications
sufficient for 100 extractions
sufficient for 100 reactions

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Tissue PCR Kits provide all the reagents necessary to rapidly extract DNA from a wide variety of cells and tissues and amplify targets of interest by PCR. A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR ReadyMix for amplification directly from the extract.

The kit contains validated protocols to extract and amplify genomic DNA from mouse-tails, hair, animal tissue, saliva, and buccal swabs. In a typical procedure, genomic DNA is extracted from a sample that has been incubated in the Tissue Preparation Solution and Extraction Solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and Extract-N-Amp PCR ReadyMix, included in the kit.
The Extract-N-Amp Tissue PCR Kits provide all the reagents necessary to rapidly extract DNA from a wide variety of cells and tissues and amplify targets of interest by direct PCR (polymerase chain reaction) . A novel extraction method eliminates the need for long enzymatic digestions or homogenization. The kit also includes a specially formulated hot start PCR ReadyMix for amplification directly from the extract.

Application

Extract-N-Amp tissue PCR kit has been used for genomic DNA extraction from mouse tail biopsies and polymerase chain reaction (PCR).

Features and Benefits

  • Fast – rapid extraction of genomic DNA for PCR in 15 minutes
  • Convenient – no long enzymatic digestions or column purification required
  • Practical – perfect for quick genomic DNA isolation for genotyping
  • Flexible – protocols available for mouse-tails, hair, animal tissue, saliva, and buccal swabs
  • Specific – Hot start antibody for highly specific PCR amplification of genomic DNA
  • Extract stable at 4 oC for at least 6 months

Components

Kits contain Tissue Preparation Solution, Extraction Solution, and Extract-N-Amp PCR Reaction Mix.

Principle

The kit contains validated protocols to extract and amplify genomic DNA from mouse-tails, hair, animal tissue, saliva, and buccal swabs. In a typical procedure, genomic DNA is extracted from a sample that has been incubated in the tissue preparation solution and extraction solution for 10 minutes at room temperature. The sample is heated to 95 °C for 3 minutes and then mixed with a third solution to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and Extract-N-Amp PCR ReadyMix, included in the kit.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyuan Ma et al.
Parasites & vectors, 13(1), 321-321 (2020-06-24)
The plant-parasitic nematode Hoplolaimus columbus is a pathogen that uses a wide range of hosts and causes substantial yield loss in agricultural fields in North America. This study describes, for the first time, the complete mitochondrial genome of H. columbus
Bhaswati Bhattacharya et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(25), 14280-14291 (2020-06-10)
In utero mammalian development relies on the establishment of the maternal-fetal exchange interface, which ensures transportation of nutrients and gases between the mother and the fetus. This exchange interface is established via development of multinucleated syncytiotrophoblast cells (SynTs) during placentation.
Xiaodong Sun et al.
Cell cycle (Georgetown, Tex.), 12(7), 1133-1141 (2013-03-09)
Parkin, an E3 ubiquitin ligase well known for its role in the pathogenesis of juvenile Parkinson disease, has been considered as a candidate tumor suppressor in certain types of cancer. It remains unknown whether parkin is involved in the development
Chelsea D Mullins et al.
PloS one, 8(6), e67941-e67941 (2013-07-05)
Activating mutations in Ras (N- and K-) are the most common point mutations found in patients with multiple myeloma (MM) and are associated with poor clinical outcome. We sought to directly examine the role of Ras activation in MM pathogenesis
James A Green et al.
BMC neuroscience, 14, 96-96 (2013-09-10)
Myelinated axons are organized into distinct subcellular and molecular regions. Without proper organization, electrical nerve conduction is delayed, resulting in detrimental physiological outcomes. One such region is the paranode where axo-glial septate junctions act as a molecular fence to separate

Articles

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Extract-N-Amp™ Tissue PCR Kit quickly extracts PCR-ready DNA from mouse tails, providing optimized PCR mix for efficient amplification.

Protocols

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

Related Content

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Overview of common techniques and downstream applications for extraction and purification of genomic DNA, plasmid DNA, and total RNA from cells, tissue, blood, viruses, and other sample types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service