Skip to Content
Merck
All Photos(3)

Key Documents

XNAP

Sigma-Aldrich

REDExtract-N-Amp Plant PCR Kit

sufficient for 100 extractions, sufficient for 100 amplifications

Synonym(s):

Plant direct PCR kit, Plant direct PCR kit with loading dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 100 amplifications
sufficient for 100 extractions
sufficient for 100 reactions

Quality Level

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

red

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The REDExtract-N-Amp Plant PCR Kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. A novel Extraction Solution eliminates the need for conventional freezing of plant tissues with liquid nitrogen, mechanical disruption, organic extraction, column purification, or precipitation of DNA. The kit also includes a PCR ReadyMix, especially formulated for amplification directly from extract. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Genomic DNA is extracted from 0.5 to 0.7 cm plant leaf disks that have been cut with a standard paper punch and simply incubated in Extraction Solution at 95 oC for 10 minutes. An equal volume of Dilution Solution is added to the extract to neutralize inhibitory substances prior to PCR. A portion of the DNA extract is then added to a PCR reaction containing primers and the REDExtract-N-Amp ReadyMix, included in the kit.
The REDExtract-N-Amp Plant PCR Kits enable the rapid detection of plant pathogens by direct PCR (polymerase chain reaction). These kits contain all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by direct PCR. The REDExtract-N-Amp PCR ReadyMix contains a dye that acts as a tracking dye and allows for convenient direct loading of PCR reactions onto agarose gels for analysis.

Application

REDExtract-N-Amp Plant PCR Kit has been used to:
  • extract genomic DNA from mycelial lawns and leaves
  • screen leaf tissue from putative transgenic potato plants
  • perform polymerase chain reaction (PCR)
  • isolate DNA from Cannabis sativa L. seed and dried fruit body gills

Features and Benefits

  • Genomic DNA for PCR in less than 15 minutes
  • A PCR ReadyMix, specially formulated for amplification directly from extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • Extract stable at 4 °C for at least 6 months
  • Single-step extraction of plant genomic DNA for PCR in less than 15 minutes
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation required
  • Specially formulated PCR ReadyMix for use with extract
  • Hot Start antibody for highly specific PCR amplification of genomic DNA
  • REDExtract-N-Amp requires no loading buffers or tracking dyes required for gel analysis
  • Compatible with high-throughput requirements for genetic analysis of plants
  • Extract stable at 4 °C for at least 6 months

Components

Kits contain Dilution solution, Extraction solution, and REDExtract-N-Amp PCR Reaction Mix.

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.

Legal Information

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.

Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
REDExtract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siobhan M Brady et al.
Molecular systems biology, 7, 459-459 (2011-01-20)
Tightly controlled gene expression is a hallmark of multicellular development and is accomplished by transcription factors (TFs) and microRNAs (miRNAs). Although many studies have focused on identifying downstream targets of these molecules, less is known about the factors that regulate
Samuel J Harrison et al.
Plant methods, 2, 19-19 (2006-11-08)
The floral dip method of transformation by immersion of inflorescences in a suspension of Agrobacterium is the method of choice for Arabidopsis transformation. The presence of a marker, usually antibiotic- or herbicide-resistance, allows identification of transformed seedlings from untransformed seedlings.
Giuseppe Forlani et al.
Forensic science international. Genetics, 52, 102464-102464 (2021-01-19)
The availability of molecular markers able to distinguish drug-type from fiber-type Cannabis sativa cultivars would allow fast and cheap analysis of any plant specimen, including seeds and leaves. Several approaches to this issue have been described, mainly using polymorphisms in
Rebeca Cobos et al.
Applied and environmental microbiology, 81(18), 6474-6483 (2015-07-15)
Grapevine trunk fungal pathogens, such as Diplodia seriata and Phaeomoniella chlamydospora, can infect plants through pruning wounds. They cause grapevine trunk diseases and are involved in grapevine decline. Accordingly, the protection of pruning wounds is crucial for the management of
Allopatric instead of parapatric divergence in an ectomycorrhizal fungus (Laccaria trichodermophora) in tropical sky-islands
Quintero-Corrales C, et al.
Fungal Ecology, 47-47 (2020)

Articles

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Protocols

REDExtract-N-Amp Plant PCR kit includes loading dye for easy gel loading after plant genomic DNA extraction.

REDExtract-N-Amp Plant PCR kit includes loading dye for easy gel loading after plant genomic DNA extraction.

REDExtract-N-Amp Plant PCR kit includes loading dye for easy gel loading after plant genomic DNA extraction.

REDExtract-N-Amp Plant PCR kit includes loading dye for easy gel loading after plant genomic DNA extraction.

Related Content

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service