Skip to Content
Merck
All Photos(1)

Key Documents

X45

Sigma-Aldrich

Triton X-45

Synonym(s):

4-(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol, Polyethylene glycol 4-tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= ~5
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

technique(s)

protein purification: suitable

transition temp

pour point ~−26 °C

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-45 is a non-ionic surfactant that has a low hydrophile–lypophile balance (HLB) value. It easily disperses in aqueous solution at room temperature. It is useful in bioseparation processes. Triton X-45 is compatible with anionic, cationic, and other nonionic surfactants and chemically stable in most acidic and alkaline solutions.

Application

Triton X-45 has been used along with tri-n-butyl phosphate (TnBP) in the treatment of human plasma for efficient inactivation of Hepatitis C virus before transfusion.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cloud point of nonionic surfactant Triton X-45 in aqueous solution.
Wang Z, et al.
Colloids and Surfaces. B, Biointerfaces, 61(1), 118-122 (2008)
Dmitrii A Komissarenko et al.
Materials (Basel, Switzerland), 11(12) (2018-11-25)
The present study demonstrates the possibility of fabricating zirconia parts with a complex shape and internal architecture using a low-cost stereolithography-based technique. One of the critical steps in ceramics stereolithography is the preparation of a photo-curable slurry with properties that
Flavia Timpu et al.
Nano letters, 17(9), 5381-5388 (2017-08-03)
We show enhanced second-harmonic generation (SHG) from a hybrid metal-dielectric nanodimer consisting of an inorganic perovskite nanoparticle of barium titanate (BaTiO
Haozhen Ren et al.
Liver international : official journal of the International Association for the Study of the Liver, 33(3), 448-458 (2013-01-11)
Hepatic tissue engineering is considered as a possible alternative to liver transplantation for end-stage liver disease. Several methods of decellularization of xenogeneic liver are available to produce three-dimensional organ scaffolds for engineering liver tissues. However, rare studies have examined and
Hirokazu Yokoyama et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 857-860 (2012-12-29)
In this report, we present a novel approach for the elucidation of the physicochemical properties of lipid membranes by isothermal titration calorimetry (ITC) to quantify the heat absorbed during the solubilization of vesicles into TritonX-100 micelles. By using large and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service