Skip to Content
Merck
All Photos(1)

Key Documents

X305

Sigma-Aldrich

Triton X-305 solution

70% in H2O

Synonym(s):

Polyethylene glycol mono(4-tert-octylphenyl) ether, Polyethylene glycol p-octylphenol ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21
Pricing and availability is not currently available.

description

non-ionic

Quality Level

mol wt

~1526.0 g/mol

concentration

70% in H2O

technique(s)

blocking: suitable
cell culture | mammalian: suitable
hybridization: suitable
immunohistochemistry: suitable
protein purification: suitable

CMC

0.65 mM

transition temp

cloud point >100 °C

HLB

17.3

SMILES string

O(CCO)c1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-305 is a non-ionic surfactant. This octylphenol ethoxylate has higher number of ethylene oxide units, increased cmc (Critical micelle concentration) and hydrophile/lipophile balance when compared to Triton X-100.[1] Triton X-305 is compatible with anionic, cationic, and other nonionic surfactants and chemically stable in most acidic and alkaline solutions.

Application

Triton X-305 has been used:
  • In the treatment of cells for immunocytochemistry[2]
  • Along with peroxotungstic acid in the preparation of tungsten trioxide films by sol-gel method[3]
  • In the dispersion polymerization of styrene with activated poly(N-vinylpyrrolidone) (PVP) for the synthesis of lanthanide-encoded polystyrene particles[4]

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubilization of phospholipid bilayers by surfactants belonging to the Triton X series: Effect of polar group size.
Partearroyo M A, et al.
Journal of Colloid and Interface Science, 178(1), 156-159 (1996)
Surface functionalization methods to enhance bioconjugation in metal-labeled polystyrene particles.
Macromolecules, 44(12), 4801-4813 (2011)
Controlling particle size and photoelectrochemical properties of nanostructured WO3 with surfactants.
Memar A, et al.
Applied Surface Science, 305, 760-767 (2014)
Thangavel Ponrasu et al.
Polymers, 13(2) (2021-01-16)
The fast-dissolving drug delivery systems (FDDDSs) are developed as nanofibers using food-grade water-soluble hydrophilic biopolymers that can disintegrate fast in the oral cavity and deliver drugs. Jelly fig polysaccharide (JFP) and pullulan were blended to prepare fast-dissolving nanofiber by electrospinning.
Hirokazu Yokoyama et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 857-860 (2012-12-29)
In this report, we present a novel approach for the elucidation of the physicochemical properties of lipid membranes by isothermal titration calorimetry (ITC) to quantify the heat absorbed during the solubilization of vesicles into TritonX-100 micelles. By using large and

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service