Skip to Content
Merck
All Photos(4)

Key Documents

X4002

Sigma-Aldrich

Xanthine

BioUltra, ≥99%

Synonym(s):

2,6-Dihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O2
CAS Number:
Molecular Weight:
152.11
Beilstein:
8733
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

product line

BioUltra

Assay

≥99%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

solubility

1 M NaOH: 0.1 M, clear, colorless to light yellow

anion traces

sulfate (SO42-): <0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O=C1NC(=O)c2nc[nH]c2N1

InChI

1S/C5H4N4O2/c10-4-2-3(7-1-6-2)8-5(11)9-4/h1H,(H3,6,7,8,9,10,11)

InChI key

LRFVTYWOQMYALW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xanthine is a purine that can be produced in the purine metabolic pathway via different precursors:
  • Guanine deamination by guanine deaminase
  • Hypoxanthine conversion by xanthine oxidoreductase
Xanthine or its derivatives are naturally present in animal organs, yeast, potatoes, coffee beans, and tea.

Application

Xanthine has been used as a substrate for the determination of xanthine oxidase (XOD) inhibitory activity.[1][2] It has also been used in the following systems:
  • yeast culture[3]
  • human cell line culture
  • recombinant viruses cloned as bacterial artificial chromosomes in Escherichia coli

Biochem/physiol Actions

Xanthine and xanthine oxidase are used to generate superoxide radicals to measure the activity of superoxide dismutase.

Features and Benefits

  • Tested for trace metal content (fuller details in Properties section and on Specification Sheet)
  • Tested for trace sulfate content (fuller details in Properties section and on Specification Sheet)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaello Castoria et al.
Phytopathology, 93(5), 564-572 (2008-10-24)
ABSTRACT We detected the generation of the reactive oxygen species (ROS) superoxide anion ( O.(-) (2)) and hydrogen peroxide (H(2)O(2)) in apple wounds 2 immediately after wounding, and assessed the relationships between (i) timely colonization of apple wounds by biocontrol
Insights from multispectral and molecular docking investigation on the xanthine oxidase inhibition by 1, 4-dicaffeoylquinic acid
Cao W, et al.
Journal of Molecular Structure, 1219, 128475-128475 null
Optimization of total phenolic contents, antioxidant, and in-vitro xanthine oxidase inhibitory activity of sunflower head
Mehmood A, et al.
CYTA-journal of Food, 16, 957-964 null
Avery G Frey et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(22), 8031-8036 (2014-05-21)
Although cells express hundreds of metalloenzymes, the mechanisms by which apoenzymes receive their metal cofactors are largely unknown. Poly(rC)-binding proteins PCBP1 and PCBP2 are multifunctional adaptor proteins that bind iron and deliver it to ferritin for storage or to prolyl
S M Forget et al.
Organic & biomolecular chemistry, 13(3), 866-875 (2014-11-20)
Cps2L, a thymidylytransferase, is the first enzyme in Streptococcus pneumoniae L-rhamnose biosynthesis and an antibacterial target. We herein report the evaluation of six sugar phosphate analogues selected to further probe Cps2L substrate tolerance. A modified continuous spectrophotometric assay was employed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service