Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Xanthine
Synonym(s):
2,6-Dihydroxypurine
Empirical Formula (Hill Notation):
C
5
H
4
N
4
O
2
CAS Number:
69-89-6
Molecular Weight:
152.11
EC Number:
200-718-6
Products
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
X7375
Xanthine,
≥99%
Expand
View Pricing
X0626
Xanthine,
≥99.5% (HPLC), purified by recrystallization
Expand
View Pricing
X4002
Xanthine,
BioUltra, ≥99%
Expand
View Pricing