Skip to Content
Merck
All Photos(7)

Key Documents

WH0000308M1

Sigma-Aldrich

Monoclonal Anti-ANXA5 antibody produced in mouse

clone 1F4-1A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ANX5, ENX2, PP4, annexin A5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F4-1A5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA5(308)

General description

The annexin A5 (ANXA5) gene is mapped to human chromosome 4q26-q28 and spans a length of 28kb containing 13 exons and 12 introns. ANXA5 is a member of the annexin family of calcium and membrane-binding proteins that have anti-coagulatory and anti-inflammatory properties. The encoded protein is 320 amino acid long and is folded into four domains, each having five α-helices, forming a superhelical globular structure.
The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa. (provided by RefSeq)

Immunogen

ANXA5 (AAH01429, 1 a.a. ~ 320 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAQVLRGTVTDFPGFDERADAETLRKAMKGLGTDEESILTLLTSRSNAQRQEISAAFKTLFGRDLLDDLKSELTGKFEKLIVALMKPSRLYDAYELKHALKGAGTNEKVLTEIIASRTPEELRAIKQVYEEEYGSSLEDDVVGDTSGYYQRMLVVLLQANRDPDAGIDEAQVEQDAQALFQAGELKWGTDEEKFITIFGTRSVSHLRKVFDKYMTISGFQIEETIDRETSGNLEQLLLAVVKSIRSIPAYLAETLYYAMKGAGTDDHTLIRVMVSRSEIDLFNIRKEFRKNFATSLYSMIKGDTSGDYKKALLLLCGEDD

Application

The annexin A5 (ANXA5) gene is mapped to human chromosome 4q26-q28 and spans a length of 28kb containing 13 exons and 12 introns. ANXA5 is a member of the annexin family of calcium and membrane-binding proteins that have anti-coagulatory and anti-inflammatory properties. The encoded protein is 320 amino acid long and is folded into four domains, each having five α-helices, forming a superhelical globular structure.

Biochem/physiol Actions

Annexins are a family of water-soluble proteins that bind to membranes and phospholipids in a calcium-dependent manner. They participate in several cellular functions, such as membrane fusion and cytoskeletal interaction. ANXA5 is a potent anticoagulant and serves as a voltage-gated calcium channel when bound to membrane in vitro. It is found to inhibit phospholipase A2 and protein kinase C.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organization of the human annexin V (ANX5) gene.
Cookson BT
Genomics, 20, 463-467 (1994)
Crystal and molecular structure of human annexin V after refinement. Implications for structure, membrane binding and ion channel formation of the annexin family of proteins.
Huber R
Journal of Molecular Biology, 223, 683-704 (1992)
The crystal and molecular structure of human annexin V, an anticoagulant protein that binds to calcium and membranes.
Huber R
The Embo Journal, 9, 3867-3874 (1990)
Karen Oliva et al.
The Journal of endocrinology, 218(2), 165-178 (2013-05-28)
Gestational diabetes mellitus (GDM) is a significant risk factor for the type 2 diabetes epidemic in many populations. Maternal adipose tissue plays a central role in the pathophysiology of GDM. Thus, the aim of this study was to determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service