Skip to Content
Merck
All Photos(5)

Key Documents

H3506

Sigma-Aldrich

Hyaluronidase from bovine testes

Type I-S, lyophilized powder, 400-1000 units/mg solid

Synonym(s):

Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type I-S

form

lyophilized powder

specific activity

400-1000 units/mg solid

mol wt

~55 kDa (four subunits of 14 kDa each)

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

UniProt accession no.

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Hyaluronidase from bovine testes has been used for the digestion of breast tissue, heart tissue and oocytes for isolation of cells.

Suitable for use in preparation of single cell suspension for sequencing.

Biochem/physiol Actions

Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression.
These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

recommended

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heikki J Nieminen et al.
Scientific reports, 7(1), 3991-3991 (2017-06-24)
Research is ongoing to develop drug therapies to manage osteoarthritis (OA) and articular cartilage (AC) injuries. However, means to deliver drug to localized AC lesions are highly limited and not clinically available. This study investigates the capability of laser ultrasound
Highly efficient gene transfer into adult ventricular myocytes by recombinant adenovirus.
Kirshenbaum LA et al.
The Journal of Clinical Investigation, 92, 381-381 (1993)
A SAGE (serial analysis of gene expression) view of breast tumor progression.
Porter DA et al.
Cancer Research, 61, 5697-5697 (2001)
Haoheng Yan et al.
The American journal of pathology, 176(3), 1421-1432 (2010-01-30)
Mutation and loss of function in p53 are common features among human breast cancers. Here we use BALB/c-Trp53+/- mice as a model to examine the sequence of events leading to mammary tumors. Mammary gland proliferation rates were similar in both
Effect of pressure at primary drying of freeze-drying mouse sperm reproduction ability and preservation potential.
Kawase Y et al.
Reproduction (Cambridge, England), 133, 841-841 (2007)

Articles

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

Use of MULTI-seq lipid-modified oligos, protocol, and troubleshooting guide for PCR Assays and Sequencing applications.

See All

Protocols

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service