Skip to Content
Merck
All Photos(1)

Documents

TP0300

Sigma-Aldrich

Total Protein Kit, Micro Lowry, Peterson′s Modification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

usage

sufficient for ~200 assays

Quality Level

General description

This method utilizes sodium dodecylsulfate, included in the Lowry reagent, to facilitate the dissolution of relatively insoluble lipoproteins. An optional deoxycholate-TCA precipitation step permits removal of interfering substances such as Tris, sucrose, EDTA, salts, etc.

Application

Total Protein Kit, Micro Lowry, Peterson′s Modification has been used to measure protein content.

Features and Benefits

  • Complete kit for protein assays
  • Most widely cited protein assay
  • Less protein to protein variation than other assays
  • Contains reagents that can remove interfering substances

Principle

An optional deoxycholate-TCA precipitation step permits removal of interfering substances such as Tris, sucrose, EDTA, salts, etc. Absorbance at 500-800 nm (typically read at 750 nm) is used to determine protein concentration from a standard curve.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective effect of carbon monoxide pre-conditioning on LPS-induced endothelial cell stress
Bernardini C, et al.
Cell Stress & Chaperones, 15(2), 219-224 (2010)
Dalia A Elgamal et al.
Molecular and cellular endocrinology, 503, 110700-110700 (2020-01-07)
Genetic and epigenetic factors contribute equally to the pathogenesis of type 1 diabetes mellitus. Sodium butyrate (NaB) has been reported to improve glucose homeostasis by modulation of the p38/ERK MAPK pathway. This work aims to evaluate the effect of NaB
Chiara Bernardini et al.
BMC complementary and alternative medicine, 18(1), 194-194 (2018-06-27)
It is widely accepted the key role of endothelium in the onset of many chronic and acute vascular and cardiovascular diseases. In the last decade, traditional compounds utilized in "folk medicine" were considered with increasing interest to discover new bioactive
Expression of alpha-gustducin and alpha-transducin, G proteins coupled with taste receptors, in boar sperm
Spinaci M, et al.
Theriogenology, 82(1), 144-151 (2014)
Alberto Elmi et al.
Animals : an open access journal from MDPI, 10(3) (2020-03-12)
The roe deer (Capreolus capreolus) represents a spontaneous model of testicular inactivation: During winter, bucks show a suspension of spermatogenesis that starts again in spring and peaks during the breeding season (July-August). The underlying mechanisms to the regulation of the

Protocols

To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.

To determine protein content, the Warburg-Christian method refers to measuring protein samples at 280 nm using a spectrophotometer.

Related Content

Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.

Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.

Products for traditional and alternative protein quantitation techniques available, including BCA, Bradford, Lowry, and more.

Protein quantification methods, reagents, and immunoassay technology for accurately measuring the protein concentrations in a variety of samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service