Skip to Content
Merck
All Photos(1)

Documents

TP0200

Sigma-Aldrich

Total Protein Kit, Micro-Lowry, Onishi & Barr Modification

sufficient for ~50 manual assays

Synonym(s):

Total Protein Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

description

wavelength 500-750nm

Quality Level

usage

sufficient for ~50 manual assays

protein detection level measuring range

0-1 mg/mL

parameter

18-26 °C temperature
200 μL sample volume

storage temp.

2-8°C

General description

The biuret and Lowry procedures are used for protein determination. The former is widely used for clinical assays. The latter, though more sensitive, is used for investigative work and is limited by poor stability of combined reagents, non-reproducibility of color, especially at low protein concentration, and nonlinear chromogenic response with protein concentration. Ohnishi and Barr modified the biuret reagent for the Lowry procedure, thereby simplifying it while improving the stability of the combined reagent.

Application

Total Protein Kit, Micro-Lowry, Onishi & Barr Modification has been used to determine total soluble protein concentrations:
  • in the dialyzates/extracts of gliadins
  • in the extracts of flour (Avena sativa and Triticum durum) and seeds (Chenopodium quinoaSalvia hispanica L.)
  • in the stored samples of rat liver microsomal fractions

Principle

Colorimetric, Endpoint.assay. Ohnishi and Barr′s modification of micro Lowry method. According to procedure, dilute biuret reagent reacts with peptide bonds to yield a purple-blue complex, the color of which is intensified by the addition of phenol reagent. Absorbance, read at 550-750 nm, is used to determine results from a standard curve.

Kit Components Also Available Separately

Product No.
Description
SDS

  • B3934Biuret reagent, protein detection level 150-1,000 μg/mL 110 mLSDS

  • Folin & Ciocalteu’s phenol reagent 5 mL

required but not provided

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Inge Everaert et al.
American journal of physiology. Renal physiology, 318(4), F1030-F1040 (2020-03-10)
Manipulation of circulating histidine-containing dipeptides (HCD) has been shown to affect the development of diabetes and early-stage diabetic nephropathy (DN). The aim of the present study was to investigate whether such interventions, which potentially alter levels of circulating HCD, also
A Kaliszewska et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 95, 89-95 (2016-07-06)
Cereal proteins are of clinical interest because of their cytotoxic and immunogenic features being associated to allergic processes and intestinal disorders. In addition to gliadins, there has been suggested an important role for non-gluten modulating factors (nGMF) on the onset
Fei-Fan Cai et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 109(8), 1090-1104 (2020-12-06)
The concept of adding inorganic fillers into hydrogels to form hydrogel nanocomposites often provides advantageous properties which can be exploited for successful 3D biofabrication. In this study, a new composite hydrogel combining oxidized alginate-gelatin (ADA-GEL) hydrogel and Laponite® nanoclay as
Method for Detecting NADPH-Cytochrome P450 Reductase in Liver Microsomal Fractions by Using Native Polyacrylamide Gel Electrophoresis and NADPH-Diaphorase Staining
Yokoyama H, et al.
American Journal of Analytical Chemistry (2013)
David Kessel
Photochemistry and photobiology, 90(5), 1211-1213 (2014-04-26)
When the initial effect of photodynamic therapy (PDT) involves mitochondrial photodamage, an early effect is loss of the mitochondrial membrane potential (ΔΨm ). Using murine hepatoma 1c1c7 cells and a photosensitizing agent known to target mitochondria, we examined loss of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service