Skip to Content
Merck
All Photos(1)

Key Documents

T2600000

Trypsin

BRP, European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trypsin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyu Zhang et al.
International journal of nanomedicine, 16, 3217-3240 (2021-05-20)
GD2 is a mainstream biomarker for neuroblastoma (NB)-targeted therapy. Current anti-GD2 therapeutics exhibit several side effects since GD2 is also expressed at low levels on normal cells. Thus, current anti-GD2 therapeutics can be compromised by the coexistence of the target
Lulin Nie et al.
Aging, 13(20), 23739-23756 (2021-10-25)
Alzheimer's disease (AD) is the most common age-related neurodegenerative disease threatening the health of the elderly, but the available therapeutic and preventive drugs remain suboptimal. Loganin, an iridoid glycoside extracted from Cornus officinalis, is reported to have anti-inflammatory and memory-enhancing
Hui Liu et al.
Evidence-based complementary and alternative medicine : eCAM, 2021, 2470182-2470182 (2022-01-11)
Silent mating type information regulation 2 homolog 1 (SIRT1) has been reported to inhibit osteoarthritic gene expression in chondrocytes. Here, efforts in this study were made to unveil the specific role of SIRT1 in the therapy of acupuncture on cartilage
Meng Wang et al.
International journal of nanomedicine, 14, 8707-8724 (2019-12-07)
The restoration and repair method in the clinic of delayed fracture healing and non-union after comminuted fractures are urgently needed to improve the prognosis of patients. The recruitment of endogenous stem cells has been considered a promising approach in bone
Mohan Liu et al.
Human mutation, 43(5), 604-612 (2022-02-20)
Male infertility has become a serious health and social problem troubling approximately 15% of couples worldwide; however, the genetic and phenotypic heterogeneity of human infertility poses a substantial obstacle to effective diagnosis and therapy. A previous study reported that heterozygous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service