Skip to Content
Merck
All Photos(1)

Key Documents

T1706

Sigma-Aldrich

TransIT-CRISPR®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51
Pricing and availability is not currently available.

application(s)

CRISPR

shipped in

wet ice

storage temp.

−20°C

General description

Accomplish highly effective transfection of CRISPR/Cas9 DNA, RNA, Cas9-gRNA RNP (ribonucleoprotein) complexes and siRNA to knock-out, modify or knock-down genes of interest in a wide variety of cell types.

Features and Benefits

  • Highly efficient, non-liposomal polymeric transfection/delivery method for use with MISSION CRISPR/Cas9 DNA, RNA, Cas9 RNP Complexes and siRNA.
  • TransIT-CRISPR is a simple, fast, effective, and versatile. One transfection reagent to deliver CRISPR-Cas9 components in a variety of formats.
  • Effective in most cell lines including difficult-to-transfect cells.

Components

0.4ml and 1ml DNA and siRNA non-liposomal, polymeric transfection reagent

Linkage

To order MISSION® CRISPR reagents, please click here

For more information and transfection protocols, please visit SigmaAldrich.com/TransITCRISPR

Other Notes

  • TransIT-CRISPR has been tested in variety of cell types including U20S and HEK 293, A549 and HeLa among others.
  • It is recommended to transfect MISSION CRISPR DNA and RNA with the TransIT-CRISPR transfection reagent.
  • Sigma-Aldrich® is the exclusive distributor of TransIT-CRISPR from Mirus Bio LLC.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
TransIT-CRISPR is a registered trademark of Mirus Bio LLC

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aitziber Buqué et al.
Nature communications, 11(1), 3819-3819 (2020-08-01)
Hormone receptor (HR)+ breast cancer (BC) causes most BC-related deaths, calling for improved therapeutic approaches. Despite expectations, immune checkpoint blockers (ICBs) are poorly active in patients with HR+ BC, in part reflecting the lack of preclinical models that recapitulate disease

Protocols

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Guaranteed PURedit™ CRISPR synthetic gRNAs and Cas9 protein offer industry-leading on-site cutting and specificity

Related Content

The largest offering of quality Cas9 proteins ensures you have the perfect CRISPR reagents for your specific experimental needs. Rigorous purification, NLS signals, increased specificity and activity ensure your gene editing experiments are efficient and simple.

The largest offering of quality Cas9 proteins ensures you have the perfect CRISPR reagents for your specific experimental needs. Rigorous purification, NLS signals, increased specificity and activity ensure your gene editing experiments are efficient and simple.

The largest offering of quality Cas9 proteins ensures you have the perfect CRISPR reagents for your specific experimental needs. Rigorous purification, NLS signals, increased specificity and activity ensure your gene editing experiments are efficient and simple.

The largest offering of quality Cas9 proteins ensures you have the perfect CRISPR reagents for your specific experimental needs. Rigorous purification, NLS signals, increased specificity and activity ensure your gene editing experiments are efficient and simple.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service