Skip to Content
Merck
All Photos(2)

Documents

STRN10

Sigma-Aldrich

Spectrum Plant Total RNA Kit

greener alternative

sufficient for 10 purifications

Synonym(s):

Plant Total RNA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 10 purifications

Quality Level

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

RNA purification: suitable

greener alternative category

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The Spectrum Plant Total RNA Kit utilizes a lysis and binding chemistry and a convenient column-based ′bind-wash-and-elute′ format to purify up to 100 μg of total RNA from 100 mg of tissue in about 30 minutes. Typical yields range from 20–60 μg.

After grinding tissue to a fine powder in liquid nitrogen, cells are lysed and cellular debris is physically and chemically separated from endogenous RNA. RNA is then bound to a column supported silica substrate and several wash steps remove remaining contaminants.

Total RNA is eluted from the column and used in typical applications, such as Northern Blots, and RT- and qRT-PCR.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry.

Features and Benefits

  • Yields up to 60 μg of pure concentrated RNA per prep
  • Efficient protocol allows for RNA purification in 30 min or less
  • Specially designed for research with difficult plant tissues

Legal Information

Spectrum is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Liver,Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhandong Cai et al.
Frontiers in plant science, 11, 724-724 (2020-06-26)
Cadmium (Cd) is a widespread pollutant that is toxic to living organisms. Previous studies have identified certain WRKY transcription factors, which confer Cd tolerance in different plant species. In the present study, we have identified 29 Cd-responsive WRKY genes in
Neeraja Cherukupalli et al.
Frontiers in plant science, 7, 1203-1203 (2016-09-02)
Andrographis paniculata is an important medicinal plant containing various bioactive terpenoids and flavonoids. Despite its importance in herbal medicine, no ready-to-use transcript sequence information of this plant is made available in the public data base, this study mainly deals with
De novo Assembly of Leaf Transcriptome in the Medicinal Plant Andrographis paniculata.
Cherukupalli N
Frontiers in Plant Science, 7, 1203-1203 (2016)
Qun Cheng et al.
Frontiers in plant science, 11, 272-272 (2020-04-08)
Soybean [Glycine max (L.) Merr.] is an important crop for oil and protein resources worldwide, and its farming is impacted by increasing soil salinity levels. In Arabidopsis the gene EARLY FLOWERING 3 (ELF3), increased salt tolerance by suppressing salt stress
Genomic approaches for interrogating the biochemistry of medicinal plant species.
Gongora-Castillo E
Methods in Enzymology, 517, 139-159 (2012)

Related Content

The Spectrum Plant Total RNA Kit incorporates a new, patent-pending lysis and binding chemistry. This kit is intended to purify total RNA from species and tissue where most Total RNA protocols fail. Without sacrificing affordability, you can purify more total RNA per isolation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service