Skip to Content
Merck
All Photos(1)

Key Documents

77627

Supelco

Poly(vinylpolypyrrolidone)

~110 μm particle size

Synonym(s):

PVPP, Polyvinylpyrrolidone, cross-linked

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164500
NACRES:
NA.21

form

lumps

Quality Level

particle size

~110 μm

application(s)

food and beverages

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinylpolypyrrolidone) is a crosslinked homopolymer form of purified vinylpyrrolidone. It is hygroscopic in nature. It has been used widely in plant hormone purification. It binds with phenolic compounds by hydrogen bonding.

Application

It was used as lysis buffer in measuring DNA concentration. It was also used in detection of recombinant proteins from leaf extracts using Surface enhanced laser desorption ionization time-of-flight mass spectrometry (SELDI-TOF MS).

Features and Benefits

Complexes with phenolics and alkaloids for their removal from plant samples, thus preventing their modification of proteins and any interference they may cause in spectrophotometric determinations of protein content. This is also reported to improve stability of enzymes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat-stress and light-stress induce different cellular pathologies in the symbiotic dinoflagellate during coral bleaching.
Downs CA
PLoS ONE, 8(12), e77173-e77173 (2013)
A SELDI-TOF MS procedure for the detection, quantitation, and preliminary characterization of low-molecular-weight recombinant proteins expressed in transgenic plants.
Badri MA
Proteomics, 9(2), 233-241 (2009)
Gianluigi Savarese et al.
Journal of the American College of Cardiology, 62(22), 2090-2099 (2013-08-21)
The purpose of this paper was to assess whether statins reduce all-cause mortality and cardiovascular (CV) events in elderly people without established CV disease. Because of population aging, prevention of CV disease in the elderly is relevant. In elderly patients
Wim-Jan P van Boven et al.
European journal of anaesthesiology, 30(11), 685-694 (2013-09-17)
Restrictive fluid management may protect organ function and improve postoperative outcome in elderly coronary artery bypass grafting (CABG) patients. We assessed organ-specific biomarker release to study the contribution of a fluid restrictive closed circuit concept to organ protection in elderly
Nneka I Comfere et al.
Journal of the American Academy of Dermatology, 69(5), 687-692 (2013-08-13)
Dermatopathology reports influence clinical management, but it is not clear to what extent comments on margin involvement of histopathologically dysplastic nevi (HDN) influence decisions about re-excision or complete excision. We sought to determine if standardized margin comments (MCs) on HDN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service